Skip to Content
Merck
All Photos(6)

Documents

10107BC

Millipore

1-2 Test® Kit

suitable for Immuno-diffusion assays for Food and Beverages applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

product name

1-2 Test®,

Agency

AOAC® 989.13 (OMA)

product line

1-2 Test®

shelf life

6‑13 mo.

packaging

pkg of 72

manufacturer/tradename

1-2 Test®
BioControl Systems 10107-72

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Salmonella spp.

General description

The 1-2 Test is a rapid qualitative method for the detection of motile Salmonella in foods, food ingredients and environmental samples. It is based on the observation of Salmonella immobilized in motility medium by polyvalent H (flagellar) antibodies. Immobilization of motile Salmonella results in development of well-defined band of cells ImmunoBand. As an AOAC Official Method of Analysis, the 1-2 Test is one of the simplest rapid methods available for the detection of Salmonella.

Application

AOAC Official Method 989.13

Features and Benefits

No equipment required to read the results

Components

  • Test Units
  • Antibody
  • Iodine-Iodide solution
  • Test trays

Principle

The assay incorporates immunodiffusion technology to provide accurate and rapid results following a simple 1 or 2 step enrichment protocol. If present,Salmonella move through the motility media as Salmonella -specific antibodies diffuse into the media from the opposite direction. The antibodies bind to the target Salmonella antigen to form a visually apparent immunoband indicating a positive test result.

Testing Method

Immunodiffusion

Legal Information

1-2 Test is a registered trademark of Merck KGaA, Darmstadt, Germany
AOAC is a registered trademark of AOAC International

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service