63003BC
FLASH® Allergen-Indicator Protein Test
BioControl, Rapid Protein Detection for HACCP Management
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41112601
NACRES:
NB.77
Recommended Products
Quality Level
product line
MVP ICON®
form
liquid
packaging
box of 100 ea
manufacturer/tradename
BioControl Systems 63003-100
FLASH®
storage condition
protect from light
application(s)
food and beverages
microbiology
detection method
colorimetric
storage temp.
room temp
Related Categories
General description
FLASH is a total protein visual test that rapidly detects protein residues left on food contact surfaces after cleaning. Protein is a difficult food residue to remove. Most allergens are proteins, so quick verification of surface hygiene helps minimize the risk of cross-contamination to allergen-free products. Using FLASH regularly also helps reduce the opportunity for biological contamination.
Application
Food contact surfaces may appear clean but can still contain traces of protein residue. Undetected, this can provide a nutrient source for harmful bacteria to grow.
Protein tests help verify cleaning effectiveness by detecting protein residues left on food contact surfaces after cleaning.
Protein is a difficult food residue to remove. And since most food allergens are proteins, regular testing can be an effective method for verifying cleaning methods, independently validated to effectively remove allergens, are consistently applied as part of a HACCP allergen management program.
Protein tests help verify cleaning effectiveness by detecting protein residues left on food contact surfaces after cleaning.
Protein is a difficult food residue to remove. And since most food allergens are proteins, regular testing can be an effective method for verifying cleaning methods, independently validated to effectively remove allergens, are consistently applied as part of a HACCP allergen management program.
Features and Benefits
Rapid test results with clear color interpretation
Verify cleaning effectiveness quickly and consistently, allowing for immediate corrective action while minimizing downtime between production change-overs.
Obtain initial room temperature results down to 20 μg within 10 minutes.
Detects protein residues including certain “Big 8” food allergens
FLASH testing can be incorporated as a standardized component of your HACCP allergen management program. And rest assured knowing you comply with SQFI guidance for verifying adequate removal of allergenic material.
Build an even more comprehensive and flexible HACCP management program.
FLASH is a perfect complement to MVP ICON ATP testing, which is a commonly used method for monitoring overall environmental hygiene. But ATP tests can′t measure protein, which is a difficult food residue to remove from surfaces even after cleaning. Used regularly, both can be incorporated into validated cleaning procedures (SSOPs) to minimize the risk of biological contamination and cross-contamination to allergen-free products.
Verify cleaning effectiveness quickly and consistently, allowing for immediate corrective action while minimizing downtime between production change-overs.
Obtain initial room temperature results down to 20 μg within 10 minutes.
Detects protein residues including certain “Big 8” food allergens
FLASH testing can be incorporated as a standardized component of your HACCP allergen management program. And rest assured knowing you comply with SQFI guidance for verifying adequate removal of allergenic material.
Build an even more comprehensive and flexible HACCP management program.
FLASH is a perfect complement to MVP ICON ATP testing, which is a commonly used method for monitoring overall environmental hygiene. But ATP tests can′t measure protein, which is a difficult food residue to remove from surfaces even after cleaning. Used regularly, both can be incorporated into validated cleaning procedures (SSOPs) to minimize the risk of biological contamination and cross-contamination to allergen-free products.
Suitability
FLASH® Allergen indicator total protein test
Testing Method
Rapid Protein Detection
Legal Information
FLASH is a registered trademark of Merck KGaA, Darmstadt, Germany
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service