Skip to Content
Merck
All Photos(1)

Key Documents

30603

Sigma-Aldrich

Potassium hydroxide

pellets, reag. Ph. Eur., ≥85%

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥85%
form:
pellets

Agency

USP/NF
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 719 °C)

Assay

≥85%

form

pellets

impurities

≤2.0% K2CO3

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

anion traces

chloride (Cl-): ≤0.02%
phosphate (PO43-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Al: ≤0.001%
Ca: ≤0.001%
Fe: ≤0.001%
Na: ≤1.0%
Ni: ≤0.0005%

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide is a strong alkali that can be used in nucleophilic substitution reactions, addition reactions, and basic hydrolysis reactions. It can also be used to catalyze aldol-type reactions.

Application

Potassium hydroxide can be used as a base:
  • In the Pd-catalyzed Heck reaction of aryl halides with alkenes.
  • To synthesize β-alkylated alcohols via pincer-Ni catalyzed Guerbet reaction.
  • For the hydroxylation of aryl or heteroarylboronic acids to corresponding phenols.
It can also be used as a catalyst for the:
  • Transesterification of triglycerides to prepare biodiesels.
  • Synthesis of β-phenylethylamines via intermolecular addition of arylamines to styrenes.
  • Claisen-Schmidt condensation reaction to synthesize chalcones.

Packaging

This product is also available in glass bottles.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel CuCl2-cryptand-[2.2. Benzo] complex: A base free and oxidant free catalyst for Ipso-Hydroxylation of aryl/heteroaryl-boronic acids in water at room temperature
Bora S, et al.
Journal of Organometallic Chemistry, 851, 52-56 (2017)
Potassium hydroxide catalyzed addition of arylamines to styrenes
Jaspers D, et al.
Synlett, 2011, 1444-1448 (2011)
Potassium Hydroxide
Ahmed F
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
A Bronsted ammonium ionic liquid-KOH two-stage catalyst for biodiesel synthesis from crude palm oil
Man Z, et al.
Industrial Crops and Products, 144-149, 41-41 (2013)
Potential N-Heterocyclic Carbene Precursors in the Palladium-Catalyzed Heck Reaction
Demir S, et al.
Heteroatom Chem., 24, 77-83 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service