Skip to Content
Merck
All Photos(1)

Key Documents

911062

Sigma-Aldrich

Potassium hydroxide

free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352320
NACRES:
NA.21

grade

ACS reagent

Agency

suitable for EPA 1621
suitable for EPA 1633
suitable for SM 5210

vapor pressure

1 mmHg ( 719 °C)

product line

Redi-Dri

Assay

≥85%

form

pellets

quality

free-flowing

impurities

≤0.001% N compounds
≤2.0% K2CO3
10-15% water

mp

361 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
Na: ≤0.05%
Ni: ≤0.001%
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide is an inorganic salt mainly used as a precursor to synthesize other potassium compounds such as potassium acetate, potassium bicarbonate and potassium carbonate. On a commercial scale, it is synthesized by the electrolysis of potassium chloride.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Dri offers a more convenient and safer way to handle the products.
Potassium hydroxide may be used in the following processes:
  • Conversion of nitriles to amide in tert-butanol.
  • Cross-coupling of aryl halides with different sulfur-, oxygen- and nitrogen-based nucleophiles in dimethyl sulfoxide.
  • As an activator for the preparation of activated carbons from corn cobs.
  • As a base for the N-alkylation of nitrogen heterocycles in polyethylene glycol-ether solvents.
  • Synthesis of (S)-1-(4-benzyl-2-thioxothiazolidin-3-yl)propan-1-one from (S)-phenylalaninol.
  • To catalyze the polymerization of propylene oxide.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Tai Li et al.
Ultrasonics sonochemistry, 9(5), 237-239 (2002-10-10)
Claisen-Schmidt condensation of acetophenone with aromatic aldehydes catalyzed by pulverized KOH and KF-Al2O3 results chalcones in 52-97% and 83-98% yields respectively in alcoholic solvent under ultrasound irradiation.
Rokas Drevinskas et al.
Optics express, 23(2), 1428-1437 (2015-04-04)
Surface texturing is demonstrated by the combination of wet etching and ultrafast laser nanostructuring of silica glass. Using potassium hydroxide (KOH) at room temperature as an etchant of laser modified glass, we show the polarization dependent linear increase in retardance
Saeid Baroutian et al.
Bioresource technology, 102(2), 1095-1102 (2010-10-05)
In this study, a novel continuous reactor has been developed to produce high quality methyl esters (biodiesel) from palm oil. A microporous TiO2/Al2O3 membrane was packed with potassium hydroxide catalyst supported on palm shell activated carbon. The central composite design
Peng Li et al.
Angewandte Chemie (International ed. in English), 54(2), 574-577 (2014-11-15)
Self-assembly of a trigonal building subunit with diaminotriazines (DAT) functional groups leads to a unique rod-packing 3D microporous hydrogen-bonded organic framework (HOF-3). This material shows permanent porosity and demonstrates highly selective separation of C2H2/CO2 at ambient temperature and pressure.
Yoshihiro Takemura et al.
Plant physiology and biochemistry : PPB, 86, 121-129 (2014-12-02)
Endodormancy is an important feature of perennial deciduous fruit trees that survive in the extreme climates brought about by seasonal variation. To acquire a comprehensive knowledge of the biochemical processes occurring just before endodormancy breaking, the buds collected in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service