Skip to Content
Merck
All Photos(2)

Documents

60037BC

Millipore

VIP® Gold Listeria spp. Kit

suitable for immunological and lateral flow essay testing in Environmental, and Food and Beverages applications

Synonym(s):

Lateral flow test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

product name

VIP® Gold Listeria, BioControl®, Lateral flow test for detection of Listeria in food and environmental samples

Agency

AOAC® (OMA)
AOAC® (PTM)
Health Canada

product line

VIP®

form

solid

shelf life

6‑12 mo.

packaging

pkg of 40

manufacturer/tradename

BioControl Systems 60037-40
VIP®

storage condition

dry at room temperature

technique(s)

lateral flow assay: suitable
microbe id | immunological: suitable

input

food(s)

application(s)

food and beverages
pathogen testing

storage temp.

room temp

suitability

Listeria spp.

General description

VIP® Gold for Listeria is a single-step visual immunoassay for the detection of Listeria. Lateral flow tests are immunoassays for detecting food pathogens with the ultimate convenience. These tests act as mini laboratory by covering the major pathogens. They also include a built-in control reaction for optimized pathogen testing. The test is designed to use with enrichment media for food and environmental pathogen detection.

Application

VIP® Gold for Listeria is a single-step visual immunoassay for the detection of Listeria designed to be used with enrichment media in food and environmental samples

Features and Benefits

  • Fast Results: Next-day results for processed foods; raw and highly contaminated foods require an additional overnight incubation.
  • Improved Lab Efficiency: VIP Gold offers an innovative multi-test format with break-away devices designed for maximum flexibility to accommodate a wide range of testing volumes.
  • Advanced Technology: The cutting-edge particle technology allows for faster sample flow, producing a clean background, clear lines, and easy-to-read positive/negative results without the need for a reader.

Components

Foil pouch containing test devices

Principle

Each device contains a proprietary reagent system, which forms a visually apparent antigen-antibody-chromogen complex if Listeria is present.

Testing Method

Visual Immunoprecipitate

Legal Information

AOAC is a registered trademark of AOAC International
BioControl is a registered trademark of BioControl Systems, Inc.
VIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Only

Product No.
Description

  • Desiccant, 14g

Kit Components Also Available Separately

Product No.
Description
SDS

  • VIP® Gold Listeria Colloid Label Pad

  • VIP® Gold Listeria Reaction Membrane

required but not provided

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service