Skip to Content
Merck
All Photos(2)

Key Documents

C26003

Sigma-Aldrich

2-Chlorobenzotrifluoride

99%

Synonym(s):

1-Chloro-2-(trifluoromethyl)benzene, 2-Chloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CF3
CAS Number:
Molecular Weight:
180.55
Beilstein:
510993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

152 °C (lit.)

density

1.379 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1Cl

InChI

1S/C7H4ClF3/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H

InChI key

DGRVQOKCSKDWIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

135.5 °F

Flash Point(C)

57.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service