Skip to Content
Merck
All Photos(2)

Documents

52186

Supelco

(±)-Naringenin

analytical standard

Synonym(s):

(±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7-Trihydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

247-250 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC=C(C=C1)C(C2)OC3=CC(O)=CC(O)=C3C2=O

InChI

1S/C15H12O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-6,13,16-18H,7H2

InChI key

FTVWIRXFELQLPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Naringenin is a naturally occurring plant bioflavonoid present in orange and grape type of citrus fruit juices, which can exhibit pharmacological properties like anti-carcinogenic, estrogenic and anti-oxidative.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza silybum thymus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Naringenin protects against cadmium-induced oxidative renal dysfunction in rats
Renugadevi.J and Prabhu M.S
Toxicology, 256, 128-134 (2009)
Plasma kinetics and urinary excretion of the flavanones naringenin and hesperetin in humans after ingestion of orange juice and grapefruit juice
Erlund I, et al.
The Journal of Nutrition, 131, 235-241 (2001)
Mohammad Ali Esmaeili et al.
Clinical and experimental pharmacology & physiology, 41(6), 416-422 (2014-04-02)
The possible protective effects of naringenin, a naturally occurring citrus flavonone, on carbon tetrachloride (CCl4 )-induced liver injury in rats and the mechanism underlying its effects were investigated. Forty rats were divided into five groups. Rats in Groups I and
Nada Oršolić et al.
European journal of nutrition, 53(5), 1217-1227 (2013-11-26)
Reactive oxygen species play a role in a number of degenerative conditions including osteoporosis. Flavonoids as phyto-oestrogens exert physiological effects against oxidative stress diseases. We developed a retinoic acid-induced bone loss model of rats to assess whether flavonoids and alendronate
Senem Kamiloglu et al.
Journal of the science of food and agriculture, 94(11), 2225-2233 (2014-01-01)
In order to investigate the effect of home processing on the bioaccessibility of health-related constituents of tomatoes, total lycopene, phenolics, flavonoids and antioxidant capacity were determined from seven different tomato products using an in vitro gastrointestinal digestion model. Additionally, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service