Skip to Content
Merck
All Photos(1)

Documents

96353

Supelco

Kaempferol

analytical standard

Synonym(s):

3,4′,5,7-Tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Robigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
Beilstein:
304401
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H

InChI key

IYRMWMYZSQPJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kaempferol is a bioflavonoid and a main component of the extract of Ginkgo biloba. It is also present in a variety of fruits and vegetables. It may serve as a potent candidate in the treatment of postmenopausal bone loss.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Chromogenic reagent for antimony in the low ppm range and for gallium and indium in the sub-ppm range.

Biochem/physiol Actions

Potent inhibitor of osteoclastic bone resorption. The effect is believed to be attributable to both the antioxidant and estrogenic activities of kaempferol.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: allium cinnamomum crataegus echinacea eucalyptus ginkgo glycyrrhiza hypericum lycium pimpinella primula sambucus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Connective Tissue Cells: Advances in Research and Application: 2011 Edition (2012)
Drugs?Advances in Research and Application: 2012 Edition (2012)
A Gohlke et al.
Journal of dairy science, 96(4), 2303-2313 (2013-02-14)
Because of their health-promoting properties, flavonoids are used in feed supplements for ruminants, although scientific evidence for their efficacy in vivo is limited. It has been shown recently that bioavailability of quercetin is low after ruminal administration in cows because
Gil Fraqueza et al.
Dalton transactions (Cambridge, England : 2003), 41(41), 12749-12758 (2012-09-13)
Recently we demonstrated that the decavanadate (V(10)) ion is a stronger Ca(2+)-ATPase inhibitor than other oxometalates, such as the isoelectronic and isostructural decaniobate ion, and the tungstate and molybdate monomer ions, and that it binds to this protein with a
J M Calderón-Montaño et al.
Mini reviews in medicinal chemistry, 11(4), 298-344 (2011-03-25)
Epidemiological studies have revealed that a diet rich in plant-derived foods has a protective effect on human health. Identifying bioactive dietary constituents is an active area of scientific investigation that may lead to new drug discovery. Kaempferol (3,5,7-trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one) is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service