Merck
  • Naringenin attenuates CCl4 -induced hepatic inflammation by the activation of an Nrf2-mediated pathway in rats.
Naringenin attenuates CCl4 -induced hepatic inflammation by the activation of an Nrf2-mediated pathway in rats.
Clinical and experimental pharmacology & physiology (2014-04-02)
Mohammad Ali Esmaeili, Mostafa Alilou
ABSTRACT
The possible protective effects of naringenin, a naturally occurring citrus flavonone, on carbon tetrachloride (CCl4 )-induced liver injury in rats and the mechanism underlying its effects were investigated. Forty rats were divided into five groups. Rats in Groups I and II served as the normal and injured liver groups, respectively; Group III rats were treated with the standard drug silymarin as a positive control; and rats in Groups IV and V (naringenin-treated groups) were administrated 50 mg/kg, p.o., naringenin for 7 days. Liver samples were collected to evaluate mRNA and protein expression, histological changes and oxidative stress. Naringenin inhibited lipid peroxidation and reduced serum levels of hepatic enzymes induced by CCl4 . In addition, naringenin increased the liver content of reduced glutathione and the activity of anti-oxidant enzymes in rats treated with CCl4 . Naringenin attenuated liver inflammation by downregulating CCl4 -induced activation of tumour necrosis factor (TNF)-α, inducible nitric oxide synthase (iNOS) and cyclo-oxygenase (COX-2) at both the protein and mRNA levels. Naringenin treatment significantly increased NF-E2-related factor 2 (Nrf2) and heme oxygenase (HO-1) expression in injured livers. In rats treated with CCl4 alone, decreases were seen in nuclear Nrf2 expression and in the mRNA levels of its target genes (e.g. HO-1, NQO1 and glutathione S-transferase alpha 3 (GST-a3)). Together, the results suggest that naringenin can protect the liver against oxidative stress, presumably by activating the nuclear translocation of Nrf2 as well as attenuating the TNF-α pathway to elicit an anti-inflammatory response in liver tissue.
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Carbon tetrachloride, analytical standard
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Carbon tetrachloride, reagent grade, 99.9%
Supelco
Carbon tetrachloride, suitable for HPLC, ≥99.9%