Skip to Content
Merck
All Photos(1)

Documents

51239

Millipore

Oxytetra Selective Supplement

suitable for microbiology

Synonym(s):

Oxytetracycline, OGYE Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9
CAS Number:
Molecular Weight:
460.43
EC Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

environmental
food and beverages
pharmaceutical

microbiology

Mode of action

protein synthesis | interferes

suitability

molds
yeasts

SMILES string

CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O9/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31)/t12-,13-,14+,17+,21-,22+/m1/s1

InChI key

IWVCMVBTMGNXQD-PXOLEDIWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Oxytetracycline is used as an antibacterial selective agent in microbiology studies . It is a broad spectrum antibiotic that is active against a wide variety of bacteria. When added to OGY-Agar (Cat. No. 75310) Oxytetracycline suppresses the growth of accompanying bacterial flora during culturing yeasts and molds.

Biochem/physiol Actions

Oxytetracycline is a protein synthesis inhibitor .

Features and Benefits

Added to OGY-Agar (Cat. No. 75310) or OGYE ChromoSelect Agar (Cat. No. 66481) Oxytetracycline suppresses the growth of accompanying bacterial flora during culturing yeasts and moulds.

Components

(per vial sufficient for 500 ml medium)
Oxytetracycline 50.0 mg

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M MIKOLAJCIK et al.
Applied microbiology, 11, 418-422 (1963-09-01)
Induction of oxytetracycline resistance in a strain of Streptococcus lactis caused this organism to display reduced acid production, salt tolerance, pyruvate synthesis, growth at alkaline pH, and a loss in ability to produce ammonia from arginine. alpha-Ketoglutaric and oxaloacetic acids
Pavel Novy et al.
The Journal of antimicrobial chemotherapy, 66(6), 1298-1300 (2011-03-23)
In this study we examined the in vitro synergistic effect of baicalin, the flavone constituent of Scutellaria spp., in combinations with oxytetracycline and tetracycline on the growth of Staphylococcus aureus. The MICs were determined by the broth microdilution method and
P R Spooner
Parasitology, 101 Pt 3, 387-393 (1990-12-01)
Oxytetracycline (OTC) significantly inhibited cytochrome c oxidase activity in bovine lymphocytes infected with Theileria parva and in uninfected mitogen-stimulated lymphocytes. The inhibitory effect was detected in vitro within 24 h of treatment with drug concentrations as low as 1 micrograms/ml.
A L Stanton et al.
Journal of dairy science, 95(9), 4950-4960 (2012-08-25)
Bovine respiratory disease complex (BRD) is a common disease in weaned dairy calves that incurs economic and welfare costs. This study was an extension of a randomized clinical trial in which a single injection of tulathromycin (TUL) or oxytetracycline (TET)
Alireza Khadem et al.
The British journal of nutrition, 112(7), 1110-1118 (2014-09-03)
The non-antibiotic anti-inflammatory theory of antimicrobial growth promoters (AGP) predicts that alternatives can be selected by simple in vitro tests. In vitro, the known AGP oxytetracycline (OTC) and a Macleaya cordata extract (MCE) had an anti-inflammatory effect with a half-maximal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service