Skip to Content
Merck
All Photos(1)

Key Documents

O0877

Sigma-Aldrich

Oxolinic acid

quinolone antibiotic

Synonym(s):

5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO5
CAS Number:
Molecular Weight:
261.23
Beilstein:
620635
EC Number:
MDL number:
UNSPSC Code:
51282935
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

solubility

0.5 M NaOH: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

CCN1C=C(C(O)=O)C(=O)c2cc3OCOc3cc12

InChI

1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)

InChI key

KYGZCKSPAKDVKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Oxolinic acid is used to study new transmissible resistance mechanisms qnrA, qnrB, qnrS, and aac(6′)Ib-cr, in Escherichia coli and Salmonella enterica. Oxolinic acid is added to culture medium for the isolation of Gardnerella vaginalis[1].

Biochem/physiol Actions

Oxolinic acid is a quinolone antibiotic. It is a DNA-gyrase (topoisomerase II) inhibitor used for studies on DNA winding and coiling and acts as a dopamine reuptake inhibitor for studies on dopaminergic neurotransmission processes.

Other Notes

5g,25g
Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Thompson
Journal of clinical microbiology, 21(5), 843-843 (1985-05-01)
Colistin-oxolinic acid medium is proposed as a selective isolation medium for Gardnerella vaginalis. The medium is effective in inhibiting staphylococci and gram-negative bacteria while allowing growth of G. vaginalis.
J Garcia de Mateos-Verchere et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 8(4), 255-259 (1999-02-03)
The antimicrobial agent oxolinic acid, injected i.p. in mice, induced a dose dependent increase in locomotor activity. This stimulation culminated at the 32 mg/kg dose and became smaller for higher doses (64-128 mg/kg). When opposed to increasing doses (50-100-200 microg/kg
Alketa Tarushi et al.
Journal of inorganic biochemistry, 103(6), 898-905 (2009-04-28)
The neutral mononuclear zinc complexes with the quinolone antibacterial drug oxolinic acid in the absence or presence of a nitrogen donor heterocyclic ligand 2,2'-bipyridine or 1,10-phenanthroline have been synthesized and characterized. The experimental data suggest that oxolinic acid is on
Quoc Tuc Dinh et al.
Talanta, 85(3), 1238-1245 (2011-08-03)
This study presents the development of an automated on-line solid phase extraction (SPE)-liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the determination of 23 antibiotics in environmental water samples. After optimisation of LC-MS/MS conditions, SPE parameters such as sorbent type, sample
Takashi Aoki et al.
Marine biotechnology (New York, N.Y.), 13(4), 629-638 (2010-04-16)
Shrimp aquaculture is one of the major foodproducing industries in the world. However, it is being impacted by several problems including diseases, antibiotic use, and environmental factors. The extent of the effects of these problems in the immune system of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service