Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxytetra Selective Supplement
Synonym(s):
Oxytetracycline
Empirical Formula (Hill Notation):
C
22
H
24
N
2
O
9
CAS Number:
79-57-2
Molecular Weight:
460.43
EC Number:
201-212-8
Products
Product Category
media supplements (1)
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Application
Microorganism Suitability
Mode of Action
Sterility
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
51239
Oxytetra Selective Supplement,
suitable for microbiology
Expand
View Pricing