Skip to Content
Merck
All Photos(1)

Key Documents

50741

Supelco

Secologanin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥88% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](C=C)[C@@H]1CC=O

InChI

1S/C17H24O10/c1-3-8-9(4-5-18)10(15(23)24-2)7-25-16(8)27-17-14(22)13(21)12(20)11(6-19)26-17/h3,5,7-9,11-14,16-17,19-22H,1,4,6H2,2H3/t8-,9+,11-,12-,13+,14-,16+,17+/m1/s1

InChI key

CSKKDSFETGLMSB-NRZPKYKESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichiro Inoue
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 125(1), 31-49 (2005-01-07)
Secologanin, a secoiridoid glucoside, is a pivotal terpenoid intermediate in the biosynthesis of biologically active monoterpenoid indole alkaloids such as reserpine, ajmaline, and vinblastine which are biosynthesized via strictosidine, an alkaloidal glucoside, formed from secologanin and tryptamine. In secologanin biosynthesis
Christie A M Peebles et al.
Biotechnology and bioengineering, 93(3), 534-540 (2005-10-22)
Among the pharmacologically important terpenoid indole alkaloids produced by Catharanthus roseus are the anti-cancer drugs vinblastine and vincristine. These two drugs are produced in small yields within the plant, which makes them expensive to produce commercially. Metabolic engineering has focused
Souvik Kusari et al.
Journal of natural products, 74(4), 764-775 (2011-02-26)
Fungal endophytes inhabit healthy tissues of all terrestrial plant taxa studied and occasionally produce host-specific compounds. We recently isolated an endophytic fungus, Fusarium solani, from Camptotheca acuminata, capable of biosynthesizing camptothecin (CPT, 1), but this capability substantially decreased on repeated
H Yamamoto et al.
Phytochemistry, 53(1), 7-12 (2000-02-03)
Secologanin synthase, an enzyme catalyzing the oxidative cleavage of the cyclopentane ring in loganin to form secologanin, was detected in microsomal preparations from cell suspension cultures of Lonicera japonica. The reaction required NADPH and molecular oxygen, and was blocked by
G Beke et al.
Chirality, 13(8), 483-487 (2001-07-24)
In the presence of the enzyme strictosidine synthase, the coupling reaction of secologanin and tryptamine is completely stereoselective and affords strictosidine with 3S configuration, exclusively. The stereoselectivity is transferred and retained in most indole alkaloids of type I in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service