Skip to Content
Merck
All Photos(1)

Key Documents

46262

Supelco

1,1,2-Trichloroethane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHCl2
CAS Number:
Molecular Weight:
133.40
Beilstein:
1731726
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.471 (lit.)

bp

110-115 °C (lit.)

density

1.435 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClCC(Cl)Cl

InChI

1S/C2H3Cl3/c3-1-2(4)5/h2H,1H2

InChI key

UBOXGVDOUJQMTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Wang et al.
International journal of phytoremediation, 13(4), 333-344 (2011-05-24)
Plant uptake is an important process in phytoremediation. The robust uptake of volatile organic compounds (VOCs) by plants offers opportunities to establish quantitative relationships between VOCs in plant tissues and in groundwater for the purpose of phytoscreening or phytomonitoring. Most
Hee-Sung Bae et al.
Systematic and applied microbiology, 29(5), 404-413 (2005-12-13)
A novel strain, designated as BL-10(T), was characterized using a polyphasic approach after isolation from groundwater contaminated by a mixture of chlorosolvents that included 1,1,2-trichloroethane, 1,2-dichloroethane, and vinyl chloride. Stain BL-10(T) is a facultatively anaerobic bacterium able to ferment glucose
Dario Frascari et al.
Applied microbiology and biotechnology, 73(2), 421-428 (2006-10-24)
The ability of a Rhodococcus aetherovorans strain, BCP1, to grow on butane and to degrade chloroform in the 0-633 microM range (0-75.5 mg l(-1)) via aerobic cometabolism was investigated by means of resting-cell assays. BCP1 degraded chloroform with a complete
Dario Frascari et al.
Biodegradation, 16(2), 147-158 (2005-02-26)
The focus of this microcosm study was to monitor the performances of 17 butane-utilizing microcosms during a long-term (100-250 days) aerobic cometabolic depletion of chloroform (CF). The depletion of the contaminant began after a lag-time variable between 0 and 23
Constantinos V Chrysikopoulos et al.
Environmental science & technology, 38(10), 2940-2945 (2004-06-24)
The impact of acoustic pressure waves on multicomponent nonaqueous phase liquid (NAPL) ganglia dissolution in water saturated columns packed with glass beads was investigated. Laboratory data from dissolution experiments with two and three component NAPL mixtures suggested that acoustic waves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service