Skip to Content
Merck
All Photos(2)

Key Documents

36530

Supelco

Chlorfluazuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H9Cl3F5N3O3
CAS Number:
Molecular Weight:
540.65
Beilstein:
8369967
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2cc(Cl)c(Oc3ncc(cc3Cl)C(F)(F)F)c(Cl)c2

InChI

1S/C20H9Cl3F5N3O3/c21-10-5-9(30-19(33)31-17(32)15-13(24)2-1-3-14(15)25)6-11(22)16(10)34-18-12(23)4-8(7-29-18)20(26,27)28/h1-7H,(H2,30,31,32,33)

InChI key

UISUNVFOGSJSKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

B L Quesada et al.
Journal of economic entomology, 87(5), 1129-1132 (1994-10-01)
Laboratory experiments were conducted to evaluate the effectiveness of chlorfluazuron an insect growth-inhibitor against Lutzomyia longipalpis (Lutz & Neiva). Second- and third-instar L. longipalpis were exposed to four different doses (0.65, 0.75, 0.80, and 0.85 mg/ml) of chlorfluazuron added in
R F Bakr et al.
Journal of the Egyptian Society of Parasitology, 19(2), 589-597 (1989-12-01)
The biological activity and cross-resistance of some IGR's, Dimilin, BAY SIR 8514 and Chlorofluzuron against susceptible and carbaryl-resistant strains of Culex pipiens were determined. The results indicated that these compounds are highly effective against the larvae of C. pipiens but
Hematological studies on white male rats exposed to some antimoulting compounds.
I G Berberian et al.
Bulletin of environmental contamination and toxicology, 43(1), 60-65 (1989-07-01)
Brenton C Peters et al.
Journal of economic entomology, 96(6), 1828-1831 (2004-02-24)
Two aspects of the Exterra Termite Interception and Baiting System (Ensystex, Fayetteville, NC) were evaluated in a field experiment using 13 termite mounds near Townsville, Australia. First, a cellulose-acetate powder containing either 0.05% wt:wt or 0.25% wt:wt chlorfluazuron (Requiem, Ensystex
Zu-hua Shi et al.
Pest management science, 60(12), 1213-1219 (2004-12-04)
The toxicities of five pesticides commonly used in vegetable fields to the larvae of the diamondback moth, Plutella xylostella (L) and its two major parasitoids, Cotesia plutellae (Kurdjumov) and Oomyzus sokolowskii (Kurdjumov), were evaluated in the laboratory using several bioassays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service