Skip to Content
Merck
All Photos(2)

Documents

45363

Supelco

Butylat

PESTANAL®, analytical standard

Synonym(s):

S-Ethyl N,N-diisobutylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NOS
CAS Number:
Molecular Weight:
217.37
Beilstein:
1770420
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCSC(=O)N(CC(C)C)CC(C)C

InChI

1S/C11H23NOS/c1-6-14-11(13)12(7-9(2)3)8-10(4)5/h9-10H,6-8H2,1-5H3

InChI key

BMTAFVWTTFSTOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Effect of sutan on the soil microflora in corn plantings].
E Iu Ulasevich et al.
Mikrobiologicheskii zhurnal, 42(1), 39-43 (1980-01-01)
Shannon M Lynch et al.
Environmental research, 109(7), 860-868 (2009-07-21)
Although limited, epidemiologic studies suggest possible associations between butylate use and cancer risk, specifically prostate cancer and non-Hodgkin lymphoma (NHL). We examined butylate use and cancer risk more broadly in the AHS, a cohort of licensed pesticide applicators in Iowa
Effects of carbofuran and the corn rhizosphere on growth of soil microorganisms.
M A Wootton et al.
Bulletin of environmental contamination and toxicology, 50(1), 49-56 (1993-01-01)
[Effects of 2,4-D butylate herbicides on thyroid gland and testis of mice].
Huai-fu Wang et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 24(6), 375-376 (2006-07-11)
R C Peffer et al.
Xenobiotica; the fate of foreign compounds in biological systems, 21(9), 1139-1152 (1991-09-01)
1. The metabolism of the thiocarbamate herbicide SUTAN (butylate) was studied after administration of single oral doses of [isobutyl-1-14C]SUTAN to male and female rats. 2. The radiolabelled dose was rapidly absorbed and excreted, with 79% of the dose excreted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service