43147
Bromide Standard for IC
TraceCERT®, 1000 mg/L bromide in water (nominal concentration)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
concentration
1000 mg/L bromide in water (nominal concentration)
technique(s)
ion chromatography: suitable
application(s)
environmental
food and beverages
format
single component solution
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Application
Bromide standard solution has been used in the determination of bromide anion present in water samples, using ion chromatography. It has also been used as a standard solution to determine the bromide anions present in samples of sea-water, using fast ion chromatography with didodecyldimethylammonium bromide coated octadecylsilyl type of column.
Linkage
Visit the IC Portal to learn more
Analysis Note
1000 mg/L bromide in water, prepared with high purity NaBr and water
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fast ion chromatography of common inorganic anions on a short ODS column permanently coated with didodecyldimethylammonium bromide
Connolly D and Paull B
Journal of Chromatography A, 953, 299-303 (2002)
Simultaneous determination of fluoride, chloride, nitrite, bromide, nitrate, phosphate and sulfate in aqueous solutions at 10? 9 to 10? 8% level by ion chromatography
Kapinus.N.E, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 800, 321-323 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service