75335
Sodium bicarbonate/Sodium carbonate concentrate
Na2CO3 100 mM and NaHCO3 100 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 10
Synonym(s):
Sodium Bicarbonate/Sodium Carbonate Solution
About This Item
Recommended Products
Quality Level
form
liquid
concentration
100 mM Na2CO3 in H2O
100 mM NaHCO3 in H2O
technique(s)
ion chromatography: suitable
suitability
application tested
General description
Application
Linkage
Preparation Note
related product
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service