Skip to Content
Merck
All Photos(1)

Key Documents

16355

Supelco

Nitric acid

0.1 M (0.1N), eluent concentrate for IC

Synonym(s):

Nitric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NB.21

Product Name

Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC

Quality Level

concentration

0.1 M HNO3 in water (0.1N)

technique(s)

ion chromatography: suitable

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-C-161:Trimethylamine N-oxide and biogenic amines in addition to standard cations inwhite wine.
Nitric acid concentrate may be used, during removal of siloxanes in biogases.

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with nitric acid and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Schweigkofler et al.
Journal of hazardous materials, 83(3), 183-196 (2001-05-12)
Methods for the removal of gaseous silicon compounds in biogases are presented. In laboratory studies, various solid adsorption materials and liquid absorption solutions were evaluated for their siloxane elimination efficiencies. Among the liquid sorbents studied, concentrated nitric acid and sulfuric
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Stijn Smulders et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 132-140 (2014-06-14)
The unique physical and chemical properties of nanomaterials have led to their increased use in many industrial applications, including as a paint additive. For example, titanium dioxide (TiO2) engineered nanoparticles (ENPs) have well-established anti-UV, self-cleaning, and air purification effects. Silver
Samantha J Snow et al.
Journal of toxicology and environmental health. Part A, 77(19), 1164-1182 (2014-08-15)
Particulate matter (PM)-associated metals can contribute to adverse cardiopulmonary effects following exposure to air pollution. The aim of this study was to investigate how variation in the composition and size of ambient PM collected from two distinct regions in Mexico
Dawei Feng et al.
Nature communications, 6, 5979-5979 (2015-01-20)
Enzymatic catalytic processes possess great potential in chemical manufacturing, including pharmaceuticals, fuel production and food processing. However, the engineering of enzymes is severely hampered due to their low operational stability and difficulty of reuse. Here, we develop a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service