Skip to Content
Merck
All Photos(2)

Documents

41447

Supelco

L-(+)-Tartaric acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E334
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.18 (vs air)

autoignition temp.

797 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

170-172 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single
Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different
Masaharu Sugiura et al.
Organic letters, 16(19), 5172-5175 (2014-09-24)
Enantioselective conjugate addition of styrylboronic acid to dienones was effectively catalyzed by an O-monoacyltartaric acid to afford monostyrylated products with good enantioselectivity. The RCM of the monostyrylated products using the Hoveyda-Grubbs II catalyst afforded optically active cyclopentenones, including a synthetic
Jianping Yang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(43), 13642-13650 (2012-09-22)
In this paper, we report a facile one-step hydrothermal method to synthesize phase-, size-, and shape-controlled carboxyl-functionalized rare-earth fluorescence upconversion phosphors by using a small-molecule binary acid, such as malonic acid, oxalic acid, succinic acid, or tartaric acid as capping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service