Skip to Content
Merck
All Photos(2)

Key Documents

41326

Supelco

Clofibrate

analytical standard

Synonym(s):

2-(4-Chlorophenoxy)-2-methylpropionic acid ethyl ester, Ethyl 2-(4-chlorophenoxy)-2-methylpropionate, Ethyl 2-(4-chlorophenoxy)isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15ClO3
CAS Number:
Molecular Weight:
242.70
Beilstein:
1913459
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.502-1.504

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1

InChI

1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3

InChI key

KNHUKKLJHYUCFP-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Mehendale
Toxicological sciences : an official journal of the Society of Toxicology, 57(2), 187-190 (2000-09-28)
The article highlighted in this issue is "Peroxisome Proliferator-Activated Receptor Alpha-Null Mice Lack Resistance to Acetaminophen Hepatotoxicity Following Clofibrate Exposure" by Chuan Chen, Gayle E. Hennig, Herbert E. Whiteley, J Christopher Corton, and José E. Manautou (pp. 338-344).
G Calais et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 22(2), 99-103 (1991-10-01)
It is generally accepted that hypoxia is a common occurrence in many experimental and human tumours and that it is a major cause of local failure after radiotherapy. Many attempts have been made over the last years to eliminate this
Clofibrate myopathy: a case report and a review of the literature.
P Rush et al.
Seminars in arthritis and rheumatism, 15(3), 226-229 (1986-02-01)
A review of studies on the mode of action of clofibrate and betabenzalbutyrate.
H J Fallon et al.
Lipids, 7(2), 106-109 (1972-02-01)
Maryam Gholitabar et al.
The Cochrane database of systematic reviews, 12, CD009017-CD009017 (2012-12-14)
There are many pathological conditions leading to an elevated unconjugated bilirubin level (hyperbilirubinaemia) in neonates. Currently the standard therapies for unconjugated hyperbilirubinaemia include phototherapy and exchange transfusion. In addition to phototherapy, clofibrate has been studied as a treatment for hyperbilirubinaemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service