Skip to Content
Merck
All Photos(1)

Documents

1.32371

Millipore

Anaerotest Strips

Test strips to indicate an anaerobic atmosphere., pkg of 50 strips per pack

Synonym(s):

Anaerobic indicator strip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308
NACRES:
NA.24

Quality Level

form

solid

packaging

pkg of 50 strips per pack

application(s)

pharmaceutical

compatibility

Anaerocult system

storage temp.

15-25°C

General description

Anaerotest strips are designed to indicate an anaerobic atmosphere via a color change on the test strip reaction zone. An anaerobic atmosphere favors the growth of anaerobic bacteria, and this strip can indicate the absence/presence of oxygen in the environment. The methylene blue dye in the absence of oxygen undergoes a chemical reduction reaction and turns colorless thereby, indicating a total anaerobic atmosphere. The reaction zone on the test strip returns to normal (blue color) upon re-introduction of oxygen in the environment.

Application

Anaerotest strips are used to determine an anaerobic atmosphere. The absence or presence of oxygen is indicated by a color change in the reaction zone of the strip.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service