Skip to Content
Merck
All Photos(1)

Documents

38490

Supelco

5,5-Dimethyl-1,3-cyclohexanedione

for HPLC derivatization, for the determination of aldehyde formaldehyde, ≥99.0%

Synonym(s):

Dimedone, Methone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H6(=O)2
CAS Number:
Molecular Weight:
140.18
Beilstein:
471489
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (GC)
≥99.0%

quality

for the determination of aldehyde formaldehyde

mp

146-148 °C (lit.)

application(s)

general analytical

SMILES string

CC1(C)CC(=O)CC(=O)C1

InChI

1S/C8H12O2/c1-8(2)4-6(9)3-7(10)5-8/h3-5H2,1-2H3

InChI key

BADXJIPKFRBFOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,5-dimethyl-1,3-cyclohexanedione (Dimedone) is basically a highly sensitive and very specific reagent used for the determination of aldehydes. It is highly advantageous as it is environment friendly, give high yields and is a simple workup procedure.

Application

5,5-dimethyl-1,3-cyclohexanedione may be used in condensation reaction with aromatic aldehyde in ethylene glycol. It may be also be used in trace analysis of aldehydes by HPLC.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesse C Patterson et al.
Cell systems, 8(2), 163-167 (2019-02-25)
Although elevated levels of reactive oxygen species (ROS) have been observed in cancer cells and cancer cells aberrantly proliferate, it is not known whether the level of reactive oxygen species and the accumulation of oxidative damage to macromolecules vary across
Trace analysis of aldehydes by reversed-phase high-performance liquid chromatography and precolumn fluorigenic labeling with 5, 5-dimethyl-1, 3-cyclohexanedione.
Kenneth M
Journal of Chromatography A, 256, 243-252 (1983)
Solid-State Condensation Reactions Between Aldehydes and 5, 5-Dimethyl-1, 3-cyclohexanedione by Grinding at Room Temperature.
Tong-shou J
Synthetic Communications, 35 (17), 2339-2345 (2005)
Condensation of aromatic aldehydes with 5, 5-dimethyl-1, 3-cyclohexanedione without catalyst.
Shujiang Tu
Synthetic Communications, 32 (19), 3063-3067 (2002)
Joseph R Burgoyne et al.
Biochemical Society transactions, 39(5), 1260-1267 (2011-09-23)
Elevated protein oxidation is a widely reported hallmark of most major diseases. Historically, this 'oxidative stress' has been considered causatively detrimental, as the protein oxidation events were interpreted simply as damage. However, recent advances have changed this antiquated view; sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service