Skip to Content
Merck
All Photos(1)

Key Documents

I2002

Sigma-Aldrich

1,3-Indandione

97%

Synonym(s):

1,3-Dioxoindan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein:
1210061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

129-132 °C (lit.)

SMILES string

O=C1CC(=O)c2ccccc12

InChI

1S/C9H6O2/c10-8-5-9(11)7-4-2-1-3-6(7)8/h1-4H,5H2

InChI key

UHKAJLSKXBADFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Diurno et al.
Bollettino della Societa italiana di biologia sperimentale, 60(1), 79-84 (1984-01-30)
In order to obtain biologically active compounds with a probable anticoagulant activity, some new products, by Michael method from indandione and 2-phenylindandione with ethyl alpha-cyano-beta-aryl-acrylates, have been prepared. The reaction of phenylindandione with methylvinylketone has been also carried out. The
[Effect of anticoagulants of the 4-hydroxycoumarin derivative group on aminotransferase activity in vitro and in vivo].
E Marchut et al.
Folia medica Cracoviensia, 22(3-4), 331-339 (1980-01-01)
I H Hall et al.
Anticancer research, 14(5A), 2053-2058 (1994-09-01)
N-Substituted indan-1.3-diones have proven to be potent cytotoxic agents effective against the growth of single cell leukemia tumors and cell lines derived from solid tumors. A number of the derivatives were active against growth of solid tumors e.g. colon, lung
Halise Inci Gul et al.
Anti-cancer agents in medicinal chemistry, 18(12), 1770-1778 (2018-05-26)
In this study, new azafluorenones, 4-(4-fluorophenyl)-2-(4-substitutedphenyl)-5Hindeno[ 1,2-b] pyridin-5-one, I1-I8 were synthesized and chemical structures were elucidated by spectral analysis. All compounds were reported for the first time here. Compounds were tested in terms of cytotoxicity. They were found as cytotoxins/anticancer
V Violet Dhayabaran et al.
Journal of fluorescence, 27(1), 135-150 (2016-10-04)
Novel bioactive complexes of Co(II), Cu(II), Ni(II) and Zn(II) metal ions with Schiff base ligand derived from histidine and 1,3-indandione were synthesized and thoroughly characterized by various analytical and spectral techniques. The biological investigations were carried out to examine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service