Skip to Content
Merck
All Photos(1)

Key Documents

H57602

Sigma-Aldrich

2-Methyl-8-quinolinol

98%

Synonym(s):

8-Hydroxyquinaldine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
Beilstein:
119194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

bp

267 °C (lit.)

mp

71-73 °C (lit.)

SMILES string

Cc1ccc2cccc(O)c2n1

InChI

1S/C10H9NO/c1-7-5-6-8-3-2-4-9(12)10(8)11-7/h2-6,12H,1H3

InChI key

NBYLBWHHTUWMER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-8-quinolinol is a methyl substituted quinolinol derivative that shows fungicidal property.[1] It can also undergo complexation with transition metal complexes.[2]

Application

2-Methyl-8-quinolinol (HqMe) may be used as a ligand for preparing bis (2-methyl-8-quinolinolato) aluminum(III)-μ-oxo-bis (2-methyl-8-quinolinolato ) aluminum (III).[2] It may also be used in the preparation of a scandium 2-methyl-8-quinolinolate complex [Sc(qMe)4(H)].[3]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Krawczyk et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-17)
One of the main factors limiting the effectiveness of many drugs is the difficulty of their delivery to their target site in the cell and achieving the desired therapeutic dose. Moreover, the accumulation of the drug in healthy tissue can
J Thomas Leonard et al.
European journal of medicinal chemistry, 43(1), 81-92 (2007-04-25)
HIV-1 integrase inhibitory activity data of styrylquinoline derivatives have been subjected to 3D-QSAR study by molecular shape analysis (MSA) technique using Cerius(2) version 4.8 software (Accelrys). For the selection of test set compounds, initially a QSAR analysis was done based
Kristiana T Santoso et al.
Bioorganic & medicinal chemistry, 27(16), 3532-3545 (2019-07-03)
A library of thirty-two quinolinequinones (QQs) with various amine substituents at the 6- and 7-positions were synthesised efficiently and in good yields for evaluation as potential anti-tuberculosis agents. Mycobacterium tuberculosis growth inhibition assays demonstrated that QQs bearing moderate length alkyl
Antifungal activity of 5-,7-, and 5,7-substituted 2-methyl-8-quinolinols.
Gershon H, et al.
Antimicrobial Agents and Chemotherapy, 1(5), 373-375 (1972)
Infrared Spectra, Density Functional Theory and Hartree-Fock Theoretical Calculations of 2-Methyl-8-quinolinol.
Arici K and Yilmaz R.
Asian Journal of Chemistry, 25(13), 7106-7106 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service