Skip to Content
Merck
All Photos(3)

Documents

A88182

Sigma-Aldrich

o-Anisidine

≥99%

Synonym(s):

2-Aminoanisole, 2-Methoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NH2
CAS Number:
Molecular Weight:
123.15
Beilstein:
386210
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

225 °C (lit.)

mp

3-6 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1N

InChI

1S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3

InChI key

VMPITZXILSNTON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Anisidine is an aromatic amine used as a building block for the production of dyes, pigments, and pharmaceuticals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D Sangamithirai et al.
Materials science & engineering. C, Materials for biological applications, 91, 512-523 (2018-07-24)
Dopamine (DA) and Folic acid (FA) are co-existing compounds in biological fluids that plays a vital role in central nervous system and human metabolism. DA is an important neurotransmitter in the brain's neural circuits and its diminution often results in
M Mazur
The European physical journal. E, Soft matter, 22(1), 67-75 (2007-03-06)
We report on chemical polymerization of two aniline derivatives - 2-methylaniline and 2-methoxyaniline -- in a two-phase water/pentane system. We have found that poly(2-methylaniline) is produced in form of an amorphous material regardless of the fact whether the monomer is
Xin-Gui Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(18), 4573-4581 (2009-03-20)
The highest Hg-ion adsorbance so far, namely up to 2063 mg g(-1), has been achieved by poly(aniline-co-5-sulfo-2-anisidine) nanosorbents. Sorption of Hg ions occurs mainly by redox and chelation mechanisms (see scheme), but also by ion exchange and physisorption.Poly(aniline (AN)-co-5-sulfo-2-anisidine (SA))
P Santhosh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(7), 1427-1439 (2003-04-26)
UV-visible spectroelectrochemical studies on copolymerization of diphenylamine (DPA) with ortho-methoxy aniline (OMA) were carried out for different feed ratios of DPA and OMA using indium tin oxide (ITO)-coated glass as working electrode. The UV-visible spectra show clear dependencies on the
Arnab Dawn et al.
The journal of physical chemistry. B, 110(37), 18291-18298 (2006-09-15)
A novel nano-biocomposite of silver and poly(o-methoxy aniline) (POMA)/DNA hybrid has been prepared by adding DNA solution to an aqueous solution of POMA (emeraldine base, EB) and AgNO(3) mixture. The mixture was aged for 10 days and was freeze-dried to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service