Skip to Content
Merck
All Photos(1)

Key Documents

46077

Supelco

Nitenpyram

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15ClN4O2
CAS Number:
Molecular Weight:
270.72
Beilstein:
8489488
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(Cc1ccc(Cl)nc1)\C(NC)=C\[N+]([O-])=O

InChI

1S/C11H15ClN4O2/c1-3-15(11(13-2)8-16(17)18)7-9-4-5-10(12)14-6-9/h4-6,8,13H,3,7H2,1-2H3/b11-8+

InChI key

CFRPSFYHXJZSBI-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitenpyram belongs to the class of neonicotinoids with widespread use in veterinary medicines and crop production. It is effective in protecting crops as it acts against piercing-sucking pests.

Find more information here: Neonicotinoids

Application

Nitenpyram may be used as a reference standard for the determination of nitenpyram in vegetables and fruits using solid phase extraction coupled with liquid chromatography-mass spectrometry (LC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>158.0 °F

Flash Point(C)

> 70.00 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veterinary Toxicology, 521-524 (2018)
Laura Carbonell-Rozas et al.
Analytical and bioanalytical chemistry, 412(24), 6231-6240 (2019-12-12)
A simple, sensitive, and efficient method has been developed for the determination of the seven neonicotinoid insecticides commercially available (imidacloprid, thiacloprid, clothianidin, thiamethoxam, acetamiprid, nitenpyram, and dinotefuran) and the main metabolite 6-chloronicotinic acid. Micellar electrokinetic chromatography (MEKC) mode was applied
Shane Denecke et al.
Insect biochemistry and molecular biology, 91, 1-9 (2017-10-24)
ABC transporters have a well-established role in drug resistance, effluxing xenobiotics from cells and tissues within the organism. More recently, research has been dedicated to understanding the role insect ABC transporters play in insecticide toxicity, but progress in understanding the
Determination of acetamiprid, imidacloprid, and nitenpyram residues in vegetables and fruits by high-performance liquid chromatography with diode-array detection.
Obana H, et al.
Journal of Agricultural and Food Chemistry, 15(60), 4464-4467 (2002)
Determination of neonicotinoid pesticide residues in vegetables and fruits with solid phase extraction and liquid chromatography mass spectrometry.
Obana H, et al.
Journal of Agricultural and Food Chemistry, 51(9), 2501-2505 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service