Skip to Content
Merck
All Photos(1)

Key Documents

37878

Supelco

Imazaquin

PESTANAL®, analytical standard

Synonym(s):

2-(4-Isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)-quinoline-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3O3
CAS Number:
Molecular Weight:
311.34
Beilstein:
5450078
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C1(C)N=C(NC1=O)c2nc3ccccc3cc2C(O)=O

InChI

1S/C17H17N3O3/c1-9(2)17(3)16(23)19-14(20-17)13-11(15(21)22)8-10-6-4-5-7-12(10)18-13/h4-9H,1-3H3,(H,21,22)(H,19,20,23)

InChI key

CABMTIJINOIHOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Nasser et al.
Chemosphere, 75(1), 20-27 (2009-01-13)
The potential role of mechanochemical processes in enhancing degradation of imazaquin by soil components is demonstrated. The investigated components include montmorillonite saturated with Na(+), Ca(2+), Cu(2+)and Al(3+), Agsorb (a commercial clay mix), birnessite and hematite. The mechanical force applied was
Chen Xuyan et al.
Journal of AOAC International, 90(2), 568-574 (2007-05-04)
A method has been developed for the quantitation of imazaquin residues in soil. The herbicide was extracted from soil with methanol-water (2 + 1, v/v) and cleaned up by strong anion-exchange solid-phase extraction cartridges. Analysis was performed by using high-performance
Jerome B Weber et al.
Journal of agricultural and food chemistry, 51(19), 5752-5759 (2003-09-04)
Aqueous batch-type sorption-desorption studies and soil column leaching studies were conducted to determine the influence of soil properties, soil and suspension pH, and ionic concentration on the retention, release, and mobility of [14C]imazaquin in Cape Fear sandy clay loam, Norfolk
Tomas Undabeytia et al.
Water research, 42(4-5), 1211-1219 (2007-10-05)
Vesicle-clay complexes in which positively charged vesicles composed of didodecyldimethylammonium bromide (DDAB) were adsorbed on montmorillonite removed efficiently anionic (sulfentrazone, imazaquin) and neutral (alachlor, atrazine) pollutants from water. These complexes (0.5% w:w) removed 92-100% of sulfentrazone, imazaquin and alachlor and
M Cade Smith et al.
Journal of environmental quality, 32(4), 1393-1404 (2003-08-23)
Nonequilibrium disc-flow techniques may better reproduce dynamic soil-pesticide interactions than traditional batch sorption studies. Batch kinetic and equilibrium experiments and dual-label thin-disc flow experiments were conducted with atrazine (6-chloro-N-ethyl-N'-isopropyl-1,3,5-triazine-2,4-diamine) and imazaquin [2-(4,5-dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl)-3-quinolinecarboxylic acid] using a Demopolis silt loam (loamy-skeletal, carbonatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service