Skip to Content
Merck
All Photos(1)

Key Documents

37877

Supelco

Imazapyr

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N3O3
CAS Number:
Molecular Weight:
261.28
Beilstein:
5442754
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C1(C)N=C(NC1=O)c2ncccc2C(O)=O

InChI

1S/C13H15N3O3/c1-7(2)13(3)12(19)15-10(16-13)9-8(11(17)18)5-4-6-14-9/h4-7H,1-3H3,(H,17,18)(H,15,16,19)

InChI key

CLQMBPJKHLGMQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazapyr is a broad spectrum non-selective herbicide, belonging to the family of imidazolinones, which can find applications in weed control management in cereals and sugar beet. Its mode of action involves the inhibition of the synthesis of branched-chain amino acids in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of the herbicide imazapyr by Fenton reactions
Kaichouh G, et al.
Environmental Chemistry Letters, 2, 31-33 (2004)
Photocatalytic degradation of imazapyr in water: comparison of activities of different supported and unsupported TiO 2-based catalysts
Pizarro.P, et al.
Catalysis Today, 101, 211-218 (2005)
Rabiatuladawiyah Ruzmi et al.
PloS one, 15(9), e0227397-e0227397 (2020-09-15)
The continuous and sole dependence on imidazolinone (IMI) herbicides for weedy rice control has led to the evolution of herbicide resistance in weedy rice populations across various countries growing IMI herbicide-resistant rice (IMI-rice), including Malaysia. A comprehensive study was conducted
Pierluigi Barone et al.
Molecular plant, 13(8), 1219-1227 (2020-06-24)
CRISPR-Cas9 is a powerful tool for generating targeted mutations and genomic deletions. However, precise gene insertion or sequence replacement remains a major hurdle before application of CRISPR-Cas9 technology is fully realized in plant breeding. Here, we report high-frequency, selectable marker-free
Hongju Ma et al.
Pest management science, 76(11), 3785-3791 (2020-05-27)
The photosystem II (PSII)-inhibiting herbicides are important for Australian farmers to control Lolium rigidum Gaud. and other weed species in trazine tolerant (TT)-canola fields. A L. rigidum population (R) collected from a TT-canola field from Western Australia showed multiple resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service