Skip to Content
Merck
All Photos(1)

Key Documents

36588

Supelco

Simazine solution

100 μg/mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Beilstein:
10895
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

CCNc1nc(Cl)nc(NCC)n1

InChI

1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)

InChI key

ODCWYMIRDDJXKW-UHFFFAOYSA-N

General description

Simazine is a systemic triazine herbicide, which can have the tendency to suppress the photosynthesis process and block the linear electron transport in light chain.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast pesticide detection inside microfluidic device with integrated optical pH, oxygen sensors and algal fluorescence
Tahirbegi BI, et al.
Biosensors And Bioelectronics, 88, 188-195 (2017)
Electrochemical sensor based on molecularly imprinted composite membrane of poly (o-aminothiophenol) with gold nanoparticles for sensitive determination of herbicide simazine in environmental samples
Zhang J, et al.
Sensors and Actuators B, Chemical, 249, 747-755 (2017)
V Morgante et al.
Journal of environmental management, 95 Suppl, S300-S305 (2011-08-02)
Simazine is an s-triazine herbicide world widely used for the control of broadleaf weeds. The influence of leaching and microorganisms on simazine attenuation in an agricultural soil long-term treated with this herbicide was studied. To elucidate the leaching potential of
Veria Addorisio et al.
Journal of hazardous materials, 196, 242-247 (2011-10-11)
Two mesoporous metal oxides, Al(2)O(3) and Fe(2)O(3), were evaluated as regards their ability to remove simazine, a highly persistent herbicide of s-triazines, using a batch equilibrium method. The effect of several experimental parameters such as pH, contact time, initial concentration
Daniel Eikel et al.
Rapid communications in mass spectrometry : RCM, 25(16), 2345-2354 (2011-07-19)
An automated surface-sampling technique called liquid extraction surface analysis (LESA), coupled with infusion nano-electrospray high-resolution mass spectrometry and tandem mass spectrometry (MS/MS), is described and applied to the qualitative determination of surface chemical residues resulting from the artificial spraying of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service