Skip to Content
Merck
All Photos(1)

Key Documents

32374

Supelco

Fenthion oxon sulfoxide

PESTANAL®, analytical standard

Synonym(s):

Dimethyl 3-methyl-4-(methylsulfinyl)phenyl phosphate, Fenoxon sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15O5PS
CAS Number:
Molecular Weight:
278.26
Beilstein:
8686237
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

O=P(OC)(OC)OC1=CC(C)=C(S(C)=O)C=C1

InChI

1S/C10H15O5PS/c1-8-7-9(5-6-10(8)17(4)12)15-16(11,13-2)14-3/h5-7H,1-4H3

InChI key

GTZCKTIZOGTWQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenthion oxon sulfoxide is a metabolite of fenthion, obtained by oxidative desulfuration. It is widely used as a pesticide in controlling insects in numerous crops. Its mode of action involves the inhibition of acetylcholinesterase.

Application

Fenthion oxon sulfoxide may be used as a reference standard in the determination of fenthion oxon sulfoxide in water samples using:
  • Liquid chromatography coupled with triple quadrupole mass spectrometry (LC–QqQ-MS) and liquid chromatography coupled with hybrid quadrupole time-of-flight mass spectrometry(LC–QTOF-MS).
  • High performance liquid chromatography with diode array detection (HPLC-DAD) and atmospheric pressure chemical ionization mass spectrometric detection (HPLC-APCI).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Pathways of Agrochemicals: Insecticides and fungicides (2007)
Combined use of liquid chromatography triple quadrupole mass spectrometry and liquid chromatography quadrupole time-of-flight mass spectrometry in systematic screening of pesticides and other contaminants in water samples
Masia.A, et al.
Analytica Chimica Acta, 761, 117-127 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service