Skip to Content
Merck
All Photos(1)

Key Documents

34127

Supelco

Patulin solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

4-Hydroxy-4H-furo[3,2-c]pyran-2(6H)-one, PAT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

OC1OCC=C2OC(=O)C=C12

InChI

1S/C7H6O4/c8-6-3-4-5(11-6)1-2-10-7(4)9/h1,3,7,9H,2H2

InChI key

ZRWPUFFVAOMMNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Patulin is a mycotoxin produced by several mold species. It has been found in fruit, especially apples, fruit juice, cider and stewed fruits. Because of its highly toxic properties, this compound has been the focus of many studies.
Patulin is an important mycotoxin produced by several species of mold, especially within PenicilliumAspergillus and Byssochlamys. It is commonly found in fruits especially apples, pears and grapes.

Application

Patulin solution may be used as an analytical reference standard for the determination of the analyte in:
  • dried apple rings using high-performance liquid chromatography with diode array detection (HPLC-DAD)(1)
  • fruit juices using high-performance liquid chromatography with diode array detection (HPLC-DAD) and gas chromatography coupled to mass spectrometry (GC-MS)(2)

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC-DAD method for the determination of patulin in dried apple rings.
Katerere RD, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 19(4), 389-392 (2008)
S Hatab et al.
Journal of applied microbiology, 112(5), 892-899 (2012-03-08)
Apples and apple products are the most notably commodities contaminated with patulin (PAT), which cause detrimental effects on human health and economic problems. The primary objective of this study was to investigate the removal of PAT contamination from apple juice
Alicia Rodríguez et al.
International journal of food microbiology, 155(1-2), 10-18 (2012-02-14)
A multiplex real-time PCR (qPCR) method to quantify aflatoxin, ochratoxin A (OTA) and patulin producing molds in foods was developed. For this, the primer pairs F/R-omt, F/R-npstr and F/R-idhtrb and the TaqMan probes, OMTprobe, NPSprobe and IDHprobe targeting the omt-1
Assunta Raiola et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(9), 3068-3072 (2012-06-13)
Apple juices and purees represent categories widely consumed by whole population and above all children. Patulin (PAT) is a mycotoxin known for its acute and chronic effects in animals. Several studies indicate there is a risk associated to the PAT
Eszter Horváth et al.
Acta biologica Hungarica, 63(3), 389-398 (2012-09-12)
The effects of the mycotoxin patulin on the thermodynamics and kinetics of the transition of bovine serum albumin (BSA) in aqueous solution were studied by Differential Scanning Calorimetry and Photoluminescence methods. Results show that in the presence of patulin, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service