Skip to Content
Merck
All Photos(1)

Key Documents

32411

Supelco

Ochratoxin B solution

~10 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO6
CAS Number:
Molecular Weight:
369.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@@H]1Cc2ccc(C(=O)N[C@@H](Cc3ccccc3)C(O)=O)c(O)c2C(=O)O1

InChI

1S/C20H19NO6/c1-11-9-13-7-8-14(17(22)16(13)20(26)27-11)18(23)21-15(19(24)25)10-12-5-3-2-4-6-12/h2-8,11,15,22H,9-10H2,1H3,(H,21,23)(H,24,25)/t11-,15+/m1/s1

InChI key

DAEYIVCTQUFNTM-ABAIWWIYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial
Ochratoxin is a mycotoxin, produced by genus Aspergillus and Penicillium. Ochratoxin producers are widely distributed in foods such as cereals. Ochratoxins are isocoumarin derivatives. Ochratoxin B is dechlorinated analog of ochratoxin B.

Application

Ochratoxin B was used as standard to investigate the degradation of ochratoxin A and B by white rot fungus during solid state fermentation of barley contaminated with ochratoxins. It may be used in displacement immunoassay for detection of ochratoxin A.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jing Qian et al.
Biosensors & bioelectronics, 166, 112443-112443 (2020-08-11)
This work is aimed to develop of a new class of versatile aptasensor to specifically detect aflatoxin B1 (AFB1) using dual-channel detection method. To achieve this objective, gold nanoparticles (AuNPs) having peroxidase-like activity and capability of promoting silver deposition were
G Engelhardt
Mycotoxin research, 18(1), 37-43 (2002-03-01)
Degradation of ochratoxin A (OTA) and B (OTB) by three selected fungi during solid state fermentation of barley contaminated with ochratoxins was compared. In presence of the soil fungusRhizopus japonicus and the white rot fungusPanerochaete chrysosporium more than 60 %
Vasilica Lates et al.
Analytical and bioanalytical chemistry, 402(9), 2861-2870 (2012-02-15)
We report here the development of a new assay for the detection of ochratoxin A (OTA) based on the use of its dechlorinated analogue, ochratoxin B (OTB), in a displacement immunoassay. OTB was immobilised on controlled-pore glass beads followed by
Tiziano Iemmi et al.
Toxins, 12(11) (2020-11-12)
Ochratoxin A (OTA) is a mycotoxin produced by Aspergillus and Penicillium, capable of contaminating several foodstuffs. OTA damages primarily the kidneys, and is suspected to be a carcinogenic substance, thus maximum levels for OTA in foodstuffs have been established in
Lebao Mao et al.
Biosensors & bioelectronics, 127, 57-63 (2018-12-30)
A rapid and ultrasensitive molecularly imprinted photoelectrochemical (MIP-PEC) sensing platform based on ITO electrode modified with GO-CdS heterojunction was prepared for ultrasensitive measure of fumonisin B1 (FB1). CdS quantum dots (QDs) were combined with a suitable amount of graphene oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service