Skip to Content
Merck
All Photos(2)

Documents

34003

Supelco

HMMNI

VETRANAL®, analytical standard

Synonym(s):

2-Hydroxymethyl-1-methyl-5-nitro-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O3
CAS Number:
Molecular Weight:
157.13
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cn1c(CO)ncc1[N+]([O-])=O

InChI

1S/C5H7N3O3/c1-7-4(3-9)6-2-5(7)8(10)11/h2,9H,3H2,1H3

InChI key

JSAQDPJIVQMBAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HMMNI is a hydroxy metabolite of nitroimidazole antibiotics, belonging to a class of veterinary drugs with therapeutic applications in the treatment and prevention of certain bacterial and protozoal diseases in poultry and swine.
HMMNI is a major metabolite of nitroimidazole drug, dimetridazole (DMZ).

Application

HMMNI may be used as a reference standard for the determination of HMMNI in porcine liver using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
HMMNI may be used as an analytical reference standard for the determination of the analyte in:
  • Different matrices of swine using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) and ultra-performance liquid chromatography coupled to electrospray tandem mass spectrometry (UPLC-ESI-MS/MS).
  • Biological & environmental matrices using liquid chromatography with ultraviolet detection (LC-UV).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical methodologies for the determination of nitroimidazole residues in biological and environmental liquid samples: a review.
Mahugo-Santana C, et al.
Analytica Chimica Acta, 665(2), 113-122 (2010)
Determination of 5-nitroimidazoles and corresponding hydroxy metabolites in swine kidney by ultra-performance liquid chromatography coupled to electrospray tandem mass spectrometry.
Xia Xi, et al.
Analytica Chimica Acta, 637(1-2), 79-86 (2009)
S Inghelbrecht et al.
Journal of veterinary pharmacology and therapeutics, 19(1), 62-67 (1996-02-01)
The anti-trichomonal efficacy and pharmacokinetics of dimetridazole were investigated in the homing pigeon (Columba livia). Dimetridazole was formulated for drinking water medication and as a prolonged-release tablet. To suppress a Trichomonas gallinae infection successfully, medicated drinking water containing dimetridazole (400
G Carignan et al.
Journal - Association of Official Analytical Chemists, 71(6), 1146-1149 (1988-11-01)
A study was conducted to monitor the elimination of dimetridazole (DMZ) and its major metabolite 2-hydroxymethyl-1-methyl-5-nitroimidazole (HMMNI) in swine plasma and tissue, using a liquid chromatographic method with electrochemical detector sensitive to 0.5 ppb. The study consisted of 2 experiments.
Determination of dimetridazole in poultry tissues and eggs using liquid chromatography-thermospray mass spectrometry.
Cannavan A and Kennedy GD
Analyst, 122(9), 963-966 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service