Skip to Content
Merck
All Photos(2)

Key Documents

34007

Supelco

Metronidazole-OH

VETRANAL®, analytical standard

Synonym(s):

1-(2-Hydroxyethyl)-2-hydroxymethyl-5-nitroimidazole, Hydroxymetronidazole, MNZOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O4
CAS Number:
Molecular Weight:
187.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OCCn1c(CO)ncc1[N+]([O-])=O

InChI

1S/C6H9N3O4/c10-2-1-8-5(4-11)7-3-6(8)9(12)13/h3,10-11H,1-2,4H2

InChI key

AEHPOYAOLCAMIU-UHFFFAOYSA-N

General description

Metronidazole-OH is a genotoxic, carcinogenic and mutagenic drug, belonging to the class of nitroimidazoles, used as a broad spectrum veterinary drug for the treatment and prevention of certain bacterial and protozoal diseases in farm animals.

Application

Metronidazole-OH may be used as a reference standard for the determination of the antibiotic, metronidazole-OH in water samples and meat matrices using ultra-high-performance liquid chromatography coupled to quadrupole-linear ion trap tandem mass spectrometry (UPLC-Qtrap-MS/MS) and ultra-performance liquid chromatography (UPLC) coupled to time of flight mass spectrometry (TOF).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Da Silva et al.
Alimentary pharmacology & therapeutics, 13(10), 1335-1341 (1999-10-30)
To evaluate the usefulness of plasma hydroxy-metronidazole/metronidazole (OH-MET/MET) ratios as a dynamic liver function test in ethanol abusers with or without liver cirrhosis. Metronidazole was administered intravenously for 20 min to healthy volunteers, and to patients with alcohol-induced, non-cirrhotic hepatopathy
S L Pendland et al.
Antimicrobial agents and chemotherapy, 38(9), 2106-2110 (1994-09-01)
Metronidazole is metabolized to two major oxidative products: an acid metabolite and a hydroxy metabolite. While the activity of the acid metabolite is negligible, the activity of the hydroxy metabolite is approximately 65% of the activity of the parent drug.
Quantitative multiresidue method for about 100 veterinary drugs in different meat matrices by sub 2-particulate high-performance liquid chromatography coupled to time of flight mass spectrometry.
Kaufmann A, et al.
Journal of Chromatography A, 1194(1), 66-79 (2008)
C S Easmon et al.
The British journal of venereal diseases, 58(4), 246-249 (1982-08-01)
The hydroxy metabolite of metronidazole was found to be more active against 21 strains of Gardnerella vaginalis than the parent compound and less affected by culture in carbon dioxide. After 400 mg oral metronidazole (Flagyl) plasma concentrations of the two
M N Muscará et al.
British journal of clinical pharmacology, 40(5), 477-480 (1995-11-01)
Metronidazole pharmacokinetics were studied in patients with different degrees of liver cirrhosis, classified according to the Child-Pugh algorithm (A, B or C, as liver disease severity increases) and in schistosomic patients. Metronidazole (500 mg) was administered i.v. as a slow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service