Skip to Content
Merck
All Photos(1)

Key Documents

36152

Supelco

Methiocarb

PESTANAL®, analytical standard

Synonym(s):

Mercaptodimethur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
Beilstein:
1881431
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1cc(C)c(SC)c(C)c1

InChI

1S/C11H15NO2S/c1-7-5-9(14-11(13)12-3)6-8(2)10(7)15-4/h5-6H,1-4H3,(H,12,13)

InChI key

YFBPRJGDJKVWAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methiocarb is a broad spectrum N-methylcarbamate pesticide widely used to control snails in orchids and is also a chemical bird repellent in fruit crops.

Application

Methiocarb may be used as an analytical reference standard for the estimation of the analyte in fruits, vegetables, food products of animal origin and honey by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Calisi et al.
Ecotoxicology and environmental safety, 72(5), 1369-1377 (2009-05-05)
Earthworms are considered convenient indicators of land use and soil fertility. Recently the use of biomarkers in earthworms has been increasingly investigated. The aim of this work was to study possible pollutant-induced morphometric alterations in Eisenia foetida granulocytes in view
Fang Tian et al.
Chemosphere, 79(6), 646-651 (2010-03-02)
The kinetics and mechanism for methiocarb (MC) degradation by aqueous ClO2 were investigated under simulated water treatment conditions. Experimental results indicate that the reaction between MC and ClO2 was of second-order overall, and the rate constant rapidly increased from 0.56
Jesper Bo Nielsen et al.
Archives of dermatological research, 297(9), 395-402 (2005-11-30)
Tea tree oil (TTO) is a complex mixture of terpene hydrocarbons. Intensive topical use of TTO in different cosmetics and investigations into its potential as an antimicrobial or anti-inflammatory agent has accentuated the need for studies on the toxicity of
Martina Blazková et al.
Biosensors & bioelectronics, 25(4), 753-758 (2009-09-22)
A simple and rapid immunochromatographic assay for a sensitive and inexpensive monitoring of methiocarb in surface water was developed using a binding inhibition format on a membrane strip. In the assay, detection reagent consisted of anti-methiocarb antibody and colloidal carbon-labelled
Ilhan Altinok et al.
Toxicologic pathology, 35(3), 405-410 (2007-04-25)
Liver, spleen, trunk kidney, gills, and brain of rainbow trout (Oncorhynchus mykiss) were examined histologically after exposure to different concentrations of methiocarb (2.5 and 3.75 mg/L) or endosulfan (0.6 and 1.3 micro g/L) for 21 days. Histological recovery was also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service