Skip to Content
Merck
All Photos(1)

Key Documents

32249-M

Sigma-Aldrich

Toluene

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39011102
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.7% (GC)
application(s):
microbiology
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

impurities

≤0.0001% thiophene
≤0.001% free acid (as HCl)
≤0.001% free alkali (as NaOH)
≤0.001% non-volatile matter
≤0.003% S-compounds (as S)
≤0.02% water (Karl Fischer)

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)
n20/D 1.496-1.498

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 ppm
Ba: ≤0.1 ppm
Bi: ≤0.1 ppm
Ca: ≤0.5 ppm
Cd: ≤0.05 ppm
Co: ≤0.02 ppm
Cr: ≤0.02 ppm
Cu: ≤0.02 ppm
Fe: ≤0.1 ppm
K: ≤0.5 ppm
Li: ≤0.1 ppm
Mg: ≤0.1 ppm
Mn: ≤0.02 ppm
Mo: ≤0.1 ppm
Na: ≤0.5 ppm
Ni: ≤0.02 ppm
Pb: ≤0.1 ppm
Sr: ≤0.1 ppm
Zn: ≤0.1 ppm

SMILES string

Cc1ccccc1

application(s)

microbiology

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a benzene analog, is widely employed as an organic solvent. Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.

Application

Toluene may be used as a solvent for the preparation of 4-arm polylactide-based (PLA) star oligomer. It was used for the activation of glass slides, for in vitro test system for allergy diagnosis.

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara I Fall et al.
Analytical chemistry, 75(3), 556-562 (2003-02-15)
The described in vitro test system for allergy diagnosis is based on microscope glass slides activated with (3-glycidyloxypropyl)trimethoxysilane. Allergen solutions are immobilized as small droplets (approximately 10 nL) on the activated glass slides with a piezoelectric arrayer. In contrast to
A Pseudomonas thrives in high concentrations of toluene.
Inoue A and Horikoshi K.
Nature, 338, 264-266 (1989)
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
T Biegert et al.
European journal of biochemistry, 238(3), 661-668 (1996-06-15)
Toluene is degraded anoxically to CO2 by the denitrifying bacterium Thauera aromatica. Toluene first becomes oxidized to benzoyl-CoA by O2-independent reactions. Benzoyl-CoA is then reduced to non-aromatic products by benzoyl-CoA reductase. We set out to study the reactions employed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service