Skip to Content
Merck
All Photos(1)

Key Documents

32087

Supelco

Cylindrospermopsin solution

10 μg/mL in H2O, PESTANAL®, analytical standard

Synonym(s):

6-[(R)-Hydroxy[(2aS,3R,4S,5aS,7R)-2,2a,3,4,5,5a,6,7-octahydro-3-methyl-4-(sulfooxy)-1H-1,8,8b-triazaacenaphthylen-7-yl]methyl]-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O7S
CAS Number:
Molecular Weight:
415.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in H2O

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1[C@H](C[C@@H]2C[C@@H](N=C3NC[C@H]1N23)[C@@H](O)C4=CC(=O)NC(=O)N4)OS(O)(=O)=O

InChI

1S/C15H21N5O7S/c1-6-10-5-16-14-17-8(13(22)9-4-12(21)19-15(23)18-9)2-7(20(10)14)3-11(6)27-28(24,25)26/h4,6-8,10-11,13,22H,2-3,5H2,1H3,(H,16,17)(H,24,25,26)(H2,18,19,21,23)/t6-,7+,8-,10-,11+,13+/m1/s1

InChI key

LHJPHMKIGRLKDR-YUQKHPEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J S Metcalf et al.
The Science of the total environment, 421-422, 118-123 (2012-03-01)
There have been few studies concerning cyanotoxins in desert environments, compared with the multitude of studies of cyanotoxins in aquatic environments. However, cyanobacteria are important primary producers in desert environments, where after seasonal rains they can grow rapidly both stabilising
V R Oliveira et al.
Toxicon : official journal of the International Society on Toxinology, 60(5), 764-772 (2012-07-04)
Cylindrospermopsin is a cyanobacterial toxin of increasing environmental importance, as it can lead to disease if orally or intravenously absorbed. However, its in vivo lung impairment has not been documented. Thus, we aimed at verifying whether cylindrospermopsin can induce lung injury
Barbara Poniedziałek et al.
Environmental toxicology and pharmacology, 34(3), 651-660 (2012-09-19)
Cylindrospermopsin (CYN) is a secondary metabolite produced by several cyanobacteria species. Its potential effect on human health includes liver, kidneys, lungs, spleen and intestine injuries. CYN can be cyto- and genotoxic to a variety of cell types. Occurrence and expansion
Daniel Gutiérrez-Praena et al.
Chemosphere, 90(3), 986-992 (2012-08-02)
Cylindrospermopsin is a cyanobacterial toxin frequently implicated in cyanobacterial blooms that is approaching an almost cosmopolitan distribution pattern. Moreover, the predominant extracellular availability of this cyanotoxin makes it particularly likely to be taken up by a variety of aquatic organisms
Sara Bogialli et al.
Environmental science & technology, 47(1), 574-583 (2012-11-22)
An extraordinary bloom of Planktothrix rubescens, which can produce microcystins (MCs), was observed in early 2009 in the Occhito basin, used even as a source of drinking water in Southern Italy. Several activities, coordinated by a task force, were implemented

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service