Skip to Content
Merck
All Photos(1)

Key Documents

N5148

Sigma-Aldrich

Nodularin

cyanotoxin

Synonym(s):

1,4,8,11,15-Pentaazacyclononadecane, cyclic peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H60N8O10
CAS Number:
Molecular Weight:
824.96
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

microbial

Quality Level

Assay

≥94% (TLC)

form

film or powder

solubility

methanol: 2 mg/mL (Further dilute to 10% (v/v) methanol. Store solutions at −20°C for up to six months.)

storage temp.

2-8°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)[C@@H](NC(=O)\C(=C\C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C41H60N8O10/c1-8-31-38(54)48-34(40(57)58)26(5)36(52)46-29(15-12-20-44-41(42)43)37(53)45-28(25(4)35(51)47-30(39(55)56)18-19-33(50)49(31)6)17-16-23(2)21-24(3)32(59-7)22-27-13-10-9-11-14-27/h8-11,13-14,16-17,21,24-26,28-30,32,34H,12,15,18-20,22H2,1-7H3,(H,45,53)(H,46,52)(H,47,51)(H,48,54)(H,55,56)(H,57,58)(H4,42,43,44)/b17-16+,23-21+,31-8-/t24-,25-,26-,28-,29-,30+,32-,34+/m0/s1

InChI key

IXBQSRWSVIBXNC-HSKGSTCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nodularin is a cyclic pentapeptide toxin produced by the cyanobacteria Nodularia spumigena.

Application

Nodularin has been used to induce oxidative stress and analyse its dynamics in flounder liver. It has also been used as standard for identification and quantification by high performance liquid chromatography (HPLC).

Biochem/physiol Actions

Potent inhibitor of protein phosphatases types 1 and 2A. Tumor promoter in experimental animal model.; at higher doses, causes massive liver hemorrhage. Tumor-promoting effect is due to increasing oxidative stress, as evidenced by reduced levels of glutathione and appearance of oxidative damage in DNA bases.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Imed Maatouk et al.
Mutation research, 564(1), 9-20 (2004-10-12)
Microcystin-LR (MCYST-LR) and nodularin (NOD) produced by cyanobacteria are potent and specific hepatotoxins. The induction of free-radical formation, reduction of glutathione levels and induction of DNA damage are three major events found in rat hepatocytes treated with these hepatotoxins. However
E Sueoka et al.
Journal of cancer research and clinical oncology, 123(8), 413-419 (1997-01-01)
Nodularin is a new liver carcinogen possessing a potent tumor-promoting activity in rat liver, mediated through inhibition of protein phosphatases 1 and 2A, and a weak initiating activity. Since we previously reported evidence that nodularin up-regulated expression of the tumor
Detection of nodularin in European flounder (Platichthys flesus) in the west coast of Sweden: Evidence of nodularin mediated oxidative stress
Persson KJ, et al.
Harmful Algae, 8(6), 832-838 (2009)
M Namikoshi et al.
Chemical research in toxicology, 6(2), 151-158 (1993-03-01)
Dihydro derivatives of nodularin (1) and microcystin-LR (4), potent cyclic peptide hepatotoxins isolated from Nodularia spumigena and Microcystis aeruginosa, respectively, were prepared by sodium borohydride reduction of the dehydroamino acid residues. The two stereoisomers of both dihydronodularin (2 and 3)
Production of the cyanotoxin nodularin?a multifactorial approach
Pattanaik B, et al.
Harmful Algae, 10(1), 30-38 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service