Skip to Content
Merck
All Photos(1)

Key Documents

M6045

Sigma-Aldrich

2-Methyl-4-isothiazolin-3-one hydrochloride

≥99%

Synonym(s):

MIT hydrochloride, Methylisothiazolinone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NOS · HCl
CAS Number:
Molecular Weight:
151.61
Beilstein:
3993054
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

Quality Level

Assay

≥99%

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].CN1SC=CC1=O

InChI

1S/C4H5NOS.ClH/c1-5-4(6)2-3-7-5;/h2-3H,1H3;1H

InChI key

SJXPQSRCFCPWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-4-isothiazolin-3-one hydrochloride (MIT HCl) is a derivative of the synthetic isothiazolinone.

Application

Methylisothiazolinone (MIT HCl) has been used:


  • as a cytotoxic substance to investigate its effect on bronchial epithelial cells (BEAS-2B cells) and its role in apoptotic cell death
  • to study the effects of tyrosine phosphorylation on focal adhesion kinase (FAK) activity in the development of neural axons and dendrites

Biochem/physiol Actions

2-Methyl-4-isothiazolin-3-one hydrochloride (MIT HCl) is a skin sensitizer known to cause contact dermatitis and allergy. It is extensively used in the cosmetic industry as a preservative. MIT HCl is a neurotoxin and its prolonged exposure may lead to neuronal death and inhibition of neurite outgrowth.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenzhen Wang et al.
Biomaterials, 113, 145-157 (2016-11-07)
As common reactive oxygen species, H2O2 is widely used for bacterial inactivation and wound disinfection. However, the concentrations used are always higher than physiological levels, which frequently result in potential toxicity to healthy tissue and even delay wound healing. Here
Siamak Javanbakht et al.
Materials science & engineering. C, Materials for biological applications, 96, 302-309 (2019-01-05)
In compression with the intravenous administration, oral delivery most commonly used due to the non-invasive nature and the fact that avoids patient pain and discomfort. By consideration this aim, ibuprofen as a model drug was loaded into two-dimensional tunnels and
Shen Du et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(17), 7408-7416 (2002-08-28)
Neurodegenerative disorders in humans may be triggered or exacerbated by exposure to occupational or environmental agents. Here, we show that a brief exposure to methylisothiazolinone, a widely used industrial and household biocide, is highly toxic to cultured neurons but not
Zhengxi Wei et al.
Toxicology letters, 338, 67-77 (2020-12-09)
Chemical-peptide conjugation is the molecular initiating event in skin sensitization. The OECD test guideline uses a high-performance liquid chromatography/ultraviolet (HPLC/UV) detection method to quantify chemical-peptide conjugation in a direct peptide reactivity assay (DPRA), which measures the depletion of two synthetic
Zhichu Xiang et al.
Biomaterials, 115, 53-64 (2016-11-27)
A novel peptide (P75) targeting EGFR and HER2 is successfully screened from a one-bead-one-compound (OBOC) library containing approximately 2 × 105 peptides built with the aid of computational simulation. In vitro and in vivo analyses show that P75 binds to human epithelial growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service