Skip to Content
Merck
All Photos(1)

Key Documents

24380

Sigma-Aldrich

4-Chloro-3,5-dimethylphenol

purum, ≥98.0% (T)

Synonym(s):

4-Chloro-3,5-xylenol, 4-Chloro-sym-m-xylenol, PCMX

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(CH3)2OH
CAS Number:
Molecular Weight:
156.61
Beilstein:
1862539
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (T)

mp

114-116 °C (lit.)
114-116 °C

solubility

methanol: soluble 1 g/10 mL, clear to faintly turbid, colorless to very faintly yellow

functional group

chloro

SMILES string

Cc1cc(O)cc(C)c1Cl

InChI

1S/C8H9ClO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,1-2H3

InChI key

OSDLLIBGSJNGJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The degradation of 4-chloro-3,5-dimethylphenol (PCMX) by Fenton-like process using nanoparticulate zero-valent iron (nZVI) and hydrogen peroxide (H2O2) was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lejin Xu et al.
Environmental engineering science, 30(6), 294-301 (2013-06-20)
Degradation of 4-chloro-3,5-dimethylphenol (PCMX) by a heterogeneous Fenton-like process using nanoparticulate zero-valent iron (nZVI) and hydrogen peroxide (H
R J W Lambert
Journal of applied microbiology, 97(4), 699-711 (2004-09-11)
To analyse population minimum inhibitory concentrations (MICs) data from clinical strains of Staphylococcus aureus and Pseudomonas aeruginosa for changes over a 10-year period and to look for correlations between the antimicrobials tested. Data from the MIC study of 256 clinical
Christine Dellanno et al.
American journal of infection control, 37(8), 649-652 (2009-08-21)
The 2003 outbreak of severe acute respiratory syndrome (SARS) infected over 8000 people and killed 774. Transmission of SARS occurred through direct and indirect contact and large droplet nuclei. The World Health Organization recommended the use of household disinfectants, which
S Messager et al.
The Journal of hospital infection, 58(2), 115-121 (2004-10-12)
An ex vivo test was adapted to mimic the in vivo conditions of testing antiseptic activity on human forearms and in the European Standard Hygienic Handwash Test (BSEN 1499). The study was to validate the ex vivo protocols using 4.8%
Diah R Hadiati et al.
The Cochrane database of systematic reviews, 9(9), CD007462-CD007462 (2012-09-14)
The risk of maternal mortality and morbidity (particularly postoperative infection) is higher for caesarean section than for vaginal birth. With the increase in caesarean section, it is important that the risks to the mother are minimised as far as possible.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service