Skip to Content
Merck
All Photos(2)

Key Documents

719439

Sigma-Aldrich

XtalFluor-E®

Synonym(s):

DAST difluorosulfinium salt, (Diethylamino)difluorosulfonium tetrafluoroborate, N,N-Diethyl-S,S-difluorosulfiliminium tetrafluoroborate, N,N-Diethylamino-S,S-difluorosulfinium tetrafluoroborate, N-(Difluoro-λ4-sulfanylidene)-N-ethyl-ethanaminium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10BF6NS
CAS Number:
Molecular Weight:
229.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

84-87 °C

functional group

amine

storage temp.

−20°C

SMILES string

F[B-](F)(F)F.CC\[N+](CC)=S(\F)F

InChI

1S/C4H10F2NS.BF4/c1-3-7(4-2)8(5)6;2-1(3,4)5/h3-4H2,1-2H3;/q+1;-1

InChI key

YLNKFQWRRIXZPJ-UHFFFAOYSA-N

General description

XtalFluor-E® is widely used in formylation, ring expansion, proto-functionalization, cyclodehydration, and dehydration.

Application

Convenient crystalline highly chemoselective deoxofluorination reagent with a broad substrate scope

Deoxofluorination reagent with a better safety profile, that doesn′t generate corrosive HF which makes it suitable for use in standard borosilicate vessels, and does not react violently with water

Reactant for:
  • Preparation of fluorodisaccharides
Triisopropylsilanethiol was recently reported by MacMillian and coworkers to be used as an organocatalyst, in tandem with Ir(III) specifically Aldrich product 688096, in photoredox catalysis. This approach directly couples a cyano-containing arene with an allylic sp3 C-H bond.

Legal Information

XtalFluor-E is a registered trademark of OmegaChem, Inc.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melinda Nonn et al.
Organic letters, 17(5), 1074-1077 (2015-02-17)
The selective introduction of fluorine onto the skeleton of an aminocyclopentane or cyclohexane carboxylate has been developed through a novel and efficient fluoride opening of an activated aziridine ring with XtalFluor-E. The reaction proceeded through a stereoselective aziridination of the
XtalFluor-E: A useful and versatile reagent in organic transformations
Mohammadkhani L and Heravi MM
Journal of Fluorine Chemistry, 225, 11-20 (2019)
Francis Beaulieu et al.
Organic letters, 11(21), 5050-5053 (2009-10-06)
Aminodifluorosulfinium tetrafluoroborate salts were found to act as efficient deoxofluorinating reagents when promoted by an exogenous fluoride source and, in most cases, exhibited greater selectivity by providing less elimination byproduct as compared to DAST and Deoxo-Fluor. Aminodifluorosulfinium tetrafluoroborates are easy
Alexandre L'heureux et al.
The Journal of organic chemistry, 75(10), 3401-3411 (2010-04-22)
Diethylaminodifluorosulfinium tetrafluoroborate (XtalFluor-E) and morpholinodifluorosulfinium tetrafluoroborate (XtalFluor-M) are crystalline fluorinating agents that are more easily handled and significantly more stable than Deoxo-Fluor, DAST, and their analogues. These reagents can be prepared in a safer and more cost-efficient manner by avoiding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service