Skip to Content
Merck
All Photos(2)

Documents

234133

Sigma-Aldrich

Quinoline Yellow

Dye content 95 %

Synonym(s):

Solvent yellow 33

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11NO2
CAS Number:
Molecular Weight:
273.29
Colour Index Number:
47000
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

description

spirit soluble

Quality Level

Assay

≥95% (HPLC)

form

powder

composition

Dye content, 95%

technique(s)

titration: suitable

mp

240 °C

solubility

chloroform: 1 mg/mL, clear to slightly hazy, yellow to very deep yellow

λmax

439 nm

Amax

1.16 at 285-291 nm in methanol at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C1C(c2ccc3ccccc3n2)C(=O)c4ccccc14

InChI

1S/C18H11NO2/c20-17-12-6-2-3-7-13(12)18(21)16(17)15-10-9-11-5-1-4-8-14(11)19-15/h1-10,16H

InChI key

IZMJMCDDWKSTTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinoline Yellow belongs to the class of quinophthalone dyes. It is generally used as a food colorant. Quinoline Yellow is anionic in nature and is used to dye wool and silk. Studies show that Quinoline Yellow is not genotoxic or carcinogenic. It is considered to be safe for human health.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heinrich Zollinger
Color Chemistry: Syntheses, Properties, and Applications of Organic Dyes and Pigments (2003)
M J Palazzolo et al.
Contact dermatitis, 9(5), 367-371 (1983-09-01)
The certified drug and cosmetic dye substance known as D & C Yellow No. 11 [2-(2-quinolyl)-1,3 indandione] was tested for its dermal sensitization potential in guinea pigs. An induction regimen consisting of a single footpad injection of Complete Freund's Adjuvant
Encyclopedia of Food Safety (2013)
Allergic contact dermatitis to D & C yellow 11 in a hair cream.
B Monk
Contact dermatitis, 17(1), 57-58 (1987-07-01)
Kristina Knutson et al.
Biotechnology progress, 20(6), 1893-1896 (2004-12-04)
A laccase-mediator system (LMS) for biobleaching was applied to a bleached chemical pulp dyed with stilbene dye Direct Yellow 11. Of mediators tested, 2,2'-azinobis(3-ethylbenzthiazoline-6-sulfonate) (ABTS) was found to be more effective than either violuric acid (VA) or N-hydroxybenzotriazole (HBT), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service