Skip to Content
Merck
All Photos(2)

Key Documents

309052

Sigma-Aldrich

Quinoline Yellow

Mixture of the mono- and disulfonic acids of Quinoline Yellow

Synonym(s):

Acid Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
47005
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Quality Level

form

powder

technique(s)

titration: suitable

solubility

H2O: soluble
spirit: soluble (23,413-3)

λmax

412 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c2nc(ccc2cc(c1)[S](=O)(=O)[O-])C3C(=O)c4c(cccc4)C3=O

InChI

1S/C18H11NO8S2.2Na/c20-17-11-3-1-2-4-12(11)18(21)15(17)13-6-5-9-7-10(28(22,23)24)8-14(16(9)19-13)29(25,26)27;;/h1-8,15H,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2

InChI key

FZUOVNMHEAPVBW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Quinoline Yellow is a yellow, water-soluble, anionic quinophthalone dye. This synthetic dye is generally used as a food additive. Its applications are also seen in the cosmetic and pharmaceutical industries. It is a chinophthalon derivative applied in the compositions of skin, lips, or body cosmetics.Studies show that Quinoline Yellow is not genotoxic or carcinogenic. It is considered to be safe for human health.

Application

Quinoline Yellow has been used as an absorber for the fabrication of the micro-phantoms.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Weisz et al.
Journal of chromatography. A, 923(1-2), 87-96 (2001-08-21)
The main components of the color additive D&C Yellow No. 10 (Quinoline Yellow, Color Index No. 47005), 2-(2-quinolinyl)-1H-indene-1,3(2H)-dione-6'-sulfonic acid (6SA) and 2-(2-quinolinyl)-1H-indene-1,3(2H)-dione-8'-sulfonic acid (8SA), were isolated from the dye mixture by pH-zone-refining counter-current chromatography (CCC) in the ion-exchange mode. These
Methyldopa-induced skin rash.
G Gidseg
Southern medical journal, 79(3), 389-389 (1986-03-01)
Karen Lau et al.
Toxicological sciences : an official journal of the Society of Toxicology, 90(1), 178-187 (2005-12-15)
Exposure to non-nutritional food additives during the critical development window has been implicated in the induction and severity of behavioral disorders such as attention deficit hyperactivity disorder (ADHD). Although the use of single food additives at their regulated concentrations is
Adrian Weisz et al.
Journal of mass spectrometry : JMS, 37(10), 1025-1033 (2002-10-11)
Several positional isomers of 2-(2-quinolinyl)-1H-indene-1,3(2H)-dione mono- and disulfonic acids prepared as reference materials for development of analytical methods involved in FDA certification of D&C Yellow No. 10 (Quinoline Yellow) were found consistently to show [MH + 14](+) ions when their
M Florian et al.
Food additives and contaminants, 19(9), 803-809 (2002-10-25)
A study of the voltammetric behaviour of the food colours brilliant blue FCF (C.I. 42090), erythrosine (C.I. 45430) and quinoline yellow (C.I. 47005) in the pH range 2-10 have been carried out by cathodic#10; stripping voltammetry. At pH 4.5 (acetate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service