Skip to Content
Merck
All Photos(1)

Documents

901596

Sigma-Aldrich

Hydrazine solution

1.0 M in ethanol

Synonym(s):

Hydrazine, Nitrogen hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2
CAS Number:
Molecular Weight:
32.05
Beilstein:
878137
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

form

liquid

concentration

1.0 M in ethanol

refractive index

n/D 1.3666

density

0.7934 g/mL

SMILES string

NN

InChI

1S/H4N2/c1-2/h1-2H2

InChI key

OAKJQQAXSVQMHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrazine promotes the cation exchange reaction for fast doping Cu ions into ZnSe nanocrystals.

Application

Valuable building block provided in ethanol for convenient handling.
Hydrazine solution may be employed for the following studies:
  • As reducing agent for the reduction of the first row (3d) transition metal complexes in aqueous solution.
  • Preparation of nanoscale reduced metal oxides such as VO2(B), Cr2O3.
  • As fuel for the direct hydrazine fuel cell (DHFC).
  • As reducing agent for the synthesis of metallic monodisperse copper nanoparticles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Vogel et al.
Fresenius' journal of analytical chemistry, 366(8), 781-791 (2001-03-03)
Hydrazine reagents are a well-known group of derivatizing agents for the determination of aldehydes and ketones in liquid and gaseous samples. Within this article, the most important hydrazine reagents are critically summarized, and their major applications in different fields, including
Nilay Hazari
Chemical Society reviews, 39(11), 4044-4056 (2010-06-24)
One of the most challenging problems in small molecule activation is the development of a homogeneous catalyst for converting dinitrogen into ammonia at ambient temperatures and atmospheric pressure. A catalytic cycle based on molybdenum that converts dinitrogen into ammonia has
D P Elder et al.
Journal of pharmaceutical and biomedical analysis, 54(5), 900-910 (2010-12-15)
This is the latest of a series of reviews focused on the analysis of genotoxic impurities. This review summarises the analytical approaches reported in the literature relating to hydrazine, hydrazines, hydrazides and hydrazones. It is intended to provide guidance for
Min Yuan et al.
Dalton transactions (Cambridge, England : 2003), (31)(31), 6078-6088 (2010-05-08)
A combination of unique solvent properties of hydrazine enables the direct dissolution of a range of metal chalcogenides at ambient temperature, rendering this an extraordinarily simple and soft synthetic approach to prepare new metal chalcogenide-based materials. The extended metal chalcogenide
Yinhu Wang et al.
Biosensors & bioelectronics, 43, 180-185 (2013-01-12)
We report a novel hydrogen peroxide (H2O2) and hydrazine sensor based on low-cost poly(vinylpyrrolidone)-protected silver nanocubes (PVP-AgNCs). The monodisperse silver nanocubes were prepared by adding a trace amount of sodium sulfide in the conventional polyol synthesis for fast reduction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service