Skip to Content
Merck
All Photos(1)

Key Documents

19270

Sigma-Aldrich

Butyric anhydride

purum, ≥97.0% (NT)

Synonym(s):

Butanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CO)2O
CAS Number:
Molecular Weight:
158.19
Beilstein:
1099474
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.45 (vs air)

Quality Level

vapor pressure

10 mmHg ( 79.5 °C)

grade

purum

Assay

≥97.0% (NT)

autoignition temp.

535 °F

expl. lim.

1.1 %, 104 °F
7.6 %, 144 °F

refractive index

n20/D 1.413 (lit.)
n20/D 1.413

bp

198-199 °C (lit.)

mp

−75-−66 °C (lit.)

solubility

alcohol: soluble (with decomposition)(lit.)
diethyl ether: soluble(lit.)
water: soluble (with decomposition)(lit.)

density

0.967 g/mL at 25 °C (lit.)

functional group

anhydride
ester

SMILES string

CCCC(=O)OC(=O)CCC

InChI

1S/C8H14O3/c1-3-5-7(9)11-8(10)6-4-2/h3-6H2,1-2H3

InChI key

YHASWHZGWUONAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Butyric anhydride was used in the synthesis of:
  • dibutyrylchitin, a lipophilic chitin diester
  • N-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-n-butyramide
  • butyric-ester derivatives of hyaluronic acid

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Coradini et al.
International journal of cancer, 81(3), 411-416 (1999-04-21)
The potential clinical utility of sodium butyrate, a natural compound known to inhibit tumor-cell growth, is hampered by the difficulty of achieving effective in-vivo concentrations. The short half-life (about 5 minutes) of sodium butyrate results in rapid metabolism and excretion.
Xiaoning Shan et al.
International journal of pharmaceutics, 590, 119884-119884 (2020-09-21)
Poly(2-methyl-2-oxazoline) (PMOZ), poly(2-propyl-2-oxazoline) (PnPOZ) and poly(2-isopropyl-2-oxazoline) (PiPOZ) were synthesized by hydrolysis of 50 kDa poly(2-ethyl-2-oxazoline) (PEOZ) and subsequent reaction of the resulting poly(ethylene imine) with acetic, butyric and isobutyric anhydrides, respectively. These polymers were characterized by proton nuclear magnetic resonance, FTIR
J J Kaylor et al.
Carbohydrate research, 331(4), 439-444 (2001-06-12)
The syntheses of four analogues of N4-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-L-asparagine are described. Activated carboxylic acids were reacted with 2-acetamido-2-deoxy-beta-D-glucopyranosylamine. n-Butyric anhydride gave N-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-n-butyramide. 3-Chloropropionic anhydride was synthesized from 3-chloropropionic acid and gave N-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-3-chloropropionamide. Equilibration of the latter with ammonium bicarbonate gave N1-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-3-aminopropionamide. Succinimidyl
Nabilah Abdul Hadi et al.
Foods (Basel, Switzerland), 9(1) (2020-01-17)
Rice and quinoa starch esters were prepared by acylation using short-chain fatty acid anhydrides with different chain lengths (acetic, propionic, and butyric anhydride). A direct stoichiometric method based on the acylation reaction was used to determine the degree of substitution
Rosamaria Capuano et al.
Bio-protocol, 7(23), e2642-e2642 (2017-12-05)
Human induced pluripotent stem cells (hiPSCs) are a promising tool in cell-based therapies for degenerative diseases. A safe application of hiPSCs in vivo, requires the detection of the presence of residual undifferentiated pluripotent cells that can potentially cause the insurgence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service