Skip to Content
Merck
All Photos(3)

Documents

239690

Sigma-Aldrich

Succinic anhydride

≥99% (GC)

Synonym(s):

Dihydro-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O3
CAS Number:
Molecular Weight:
100.07
Beilstein:
108441
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.5 (vs air)

vapor pressure

1 mmHg ( 92 °C)

Assay

≥99% (GC)

form

solid

bp

261 °C (lit.)

mp

118-120 °C (lit.)

SMILES string

O=C1CCC(=O)O1

InChI

1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2

InChI key

RINCXYDBBGOEEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Succinic anhydride is a cyclic anhydride of succinic acid with a five-membered ring. It is widely used in the manufacture of polymeric materials, pharmaceuticals, agrochemicals, dyes, surface-active agents, and organic flame-retardant materials. It also finds application as a cross-linking agent in ion-exchange membranes and a curing agent for epoxy resins.

Application

Succinic anhydride can be used:
  • In the preparation of covalently cross-linked oxidized-alginate/N-succinyl-chitosan hydrogels, as injectable systems towards tissue engineering.
  • In preparing functionalized oxide surfaces on a chip.
  • As a starting material to synthesize polyesters in presence of a metal triflate as a catalyst.
  • As a cross-linking agent to enhance the mechanicalproperties and antimicrobial activities of zein films.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

314.6 °F

Flash Point(C)

157 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Allan D Rogalsky et al.
Journal of biomedical materials research. Part A, 99(3), 367-375 (2011-10-25)
Novel injectable hydrogels have been formed at 37°C under physiological pH using a polymer-polymer crosslinking reaction. Three different formulations were tested. After 1-day cure time at body temperature, the elastic modulus of unswollen samples ranged between 3 and 5 kPa
Sang Hyun Baek et al.
Analytical chemistry, 81(18), 7737-7742 (2009-08-12)
We present a novel dielectrophoretic technique that can be used to characterize molecular interactions inside a microfluidic device. Our approach allows functionalized beads which are initially at rest on a functionalized surface to be pulled away from the surface by
Eugenio Calandrini et al.
Optics express, 27(18), 25912-25919 (2019-09-13)
Nanoporous gold can be exploited as plasmonic material for enhanced spectroscopy both in the visible and in the near-infrared spectral regions. In particular, the peculiar morphology of such a substrate leads to a higher field confinement with respect to conventional
Yi Zhou et al.
Journal of nanobiotechnology, 15(1), 87-87 (2017-11-29)
Chemotherapeutic drugs used for cancer therapy frequently encounter multiple-drug resistance (MDR). Nanoscale carriers that can target tumors to accumulate and release drugs intracellularly have the greatest potential for overcoming MDR. Glucose transporter-1 (GLUT-1) and glutathione (GSH) overexpression in cancer cells
Yaroslav A Grechkin et al.
Biomedicines, 8(1) (2020-01-17)
DNA aptamers have many benefits for cell imaging, such as high affinity and specificity, easiness of chemical functionalization, and low cost of production. Among known aptamers, Sgc8-aptamer was selected against acute lymphoblastic leukemia cells with a dissociation constant in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service