Skip to Content
Merck
All Photos(1)

Key Documents

176044

Sigma-Aldrich

Naphthalene-d8

99 atom % D, ≥98% (CP)

Synonym(s):

1,2,3,4,5,6,7,8-Octadeuterionaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D8
CAS Number:
Molecular Weight:
136.22
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor density

4.4 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 25 °C)

isotopic purity

99 atom % D

Assay

≥98% (CP)

form

powder

expl. lim.

5.9 %

bp

218 °C (lit.)

mp

80-82 °C (lit.)

mass shift

M+8

storage temp.

room temp

SMILES string

[2H]c1c([2H])c([2H])c2c([2H])c([2H])c([2H])c([2H])c2c1[2H]

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

UFWIBTONFRDIAS-PGRXLJNUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Naphthalene-d8, isotopically labeled analogue of naphthalene.

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiabao Xu et al.
Analytical chemistry, 89(24), 13305-13312 (2017-11-15)
We illustrate that single-cell Raman microspectroscopy, coupled with deuterium isotope probing (Raman-DIP), provides a culture-independent and nondestructive approach to probe metabolic pathways of carbon substrates at the single-cell level. We found a distinguishable C-D vibration band at 2070-2300 cm-1 in
Polycyclic Aromat. Compd., 55-55 (1993)
Célia A Alves et al.
Environmental science and pollution research international, 22(15), 11526-11542 (2015-04-02)
The main purpose of this work was to evaluate the chemical composition of particulate matter (PM) emitted by eight different light-duty vehicles. Exhaust samples from petrol and diesel cars (Euro 3 to Euro 5) were collected in a chassis dynamometer
Amanda Kowalsick et al.
Journal of chromatography. A, 1370, 230-239 (2014-12-03)
Seasonal variation in tea (Camellia sinensis (L.) Kuntze; Theaceae) chemistry was investigated using automated sequential, multidimensional gas chromatography/mass spectrometry (GC-GC/MS). Metabolite libraries were produced for teas harvested from the Bulang Mountains in Yunnan, China before and after the onset of
Chen-Chou Wu et al.
Environmental science & technology, 49(17), 10607-10615 (2015-08-11)
Barbecuing or charcoal-grilling has become part of popular outdoor recreational activities nowadays; however, potential human health hazards through outdoor exposure to barbecue fumes have yet to be adequately quantified. To fill this knowledge gap, atmospheric size-fractioned particle and gaseous samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service