Skip to Content
Merck
All Photos(1)

Key Documents

15136

Supelco

Bisphenol A bis(3-chloro-2-hydroxypropyl) ether

analytical standard

Synonym(s):

2,2-Bis[4-(3-chloro-2-hydroxypropoxy)phenyl]propane, BADGE-2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26Cl2O4
CAS Number:
Molecular Weight:
413.33
Beilstein:
2302053
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)(c1ccc(OCC(O)CCl)cc1)c2ccc(OCC(O)CCl)cc2

InChI

1S/C21H26Cl2O4/c1-21(2,15-3-7-19(8-4-15)26-13-17(24)11-22)16-5-9-20(10-6-16)27-14-18(25)12-23/h3-10,17-18,24-25H,11-14H2,1-2H3

InChI key

PTCFDJRJOGPUFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Standard for determining toxic monomers released from polymers of the inner coating of cans[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographia, 34, 67-67 (1992)
Natalia Jatkowska et al.
The Science of the total environment, 767, 144286-144286 (2021-01-12)
Ecosystems are facing increased pressure due to the emission of many classes of emerging contaminants. However, very little is known about the interactions of these pollutants, such as bisphenols (BPs), plasticizers or pharmaceuticals. By employing bioluminescent bacteria (Microtox assay), we
Natchanun Leepipatpiboon et al.
Journal of chromatography. A, 1073(1-2), 331-339 (2005-05-25)
A gradient reversed-phase liquid chromatography with fluorescence detection method for simultaneous identification and quantification of bisphenol-A-diglycidyl ether (BADGE), bisphenol-F-diglycidyl ether (BFDGE), and their 10 derivatives in food matrixes was developed and validated for the analysis of oil-in-water- and aqueous-based foodstuffs.
Olga Pardo et al.
Journal of chromatography. A, 1107(1-2), 70-78 (2005-12-27)
A new confirmatory method for simultaneous determination of bisphenol diglycidyl ether residues (BADGE, BADGE.H(2)O, BADGE.2H(2)O, BADGE.H(2)O.HCl, BADGE.HCl, BADGE.2HCl, BFDGE and BFDGE.2HCl) from canned food has been developed. The proposed method includes extraction by pressurized liquid extraction (PLE) followed by liquid-liquid
Amira M El-Kosasy et al.
Journal of chromatographic science, 56(10), 920-932 (2018-07-10)
Gradient reversed-phase high-performance liquid chromatography with photodiode array detection was used for separation, detection and quantification of bisphenol-A-diglycidyl ether (BADGE) and some of its reaction products, namely, BADGE·HCl·H2O, BADGE·H2O and BADGE·2HCl in pure form and in canned foods, where canned

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service