Skip to Content
Merck
All Photos(3)

Documents

73417

Supelco

Bisphenol A (2,3-dihydroxypropyl) glycidyl ether

analytical standard

Synonym(s):

2-[4-(2,3-Dihydroxypropyloxy)phenyl]-2-[4-(glycidyloxy)phenyl]propane, BADGE-H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Beilstein:
8276468
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(c1ccc(OCC(O)CO)cc1)c2ccc(OCC3CO3)cc2

InChI

1S/C21H26O5/c1-21(2,15-3-7-18(8-4-15)24-12-17(23)11-22)16-5-9-19(10-6-16)25-13-20-14-26-20/h3-10,17,20,22-23H,11-14H2,1-2H3

InChI key

NBLIPZBCGXIEFO-UHFFFAOYSA-N

General description

Bisphenol A (2,3-dihydroxypropyl) glycidyl ether can be obtained as a product of the reaction of bisphenol A with epichlorohydrin.It is generally used as a substrate to prepare epoxy resins, which is used in coatings and adhesives.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Standard for determination of toxic BADGE and derivatives in canned food

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bisphenols, Benzophenones, and Bisphenol A Diglycidyl Ethers in Textiles and Infant Clothing
Xue J, et al.
Environmental Science & Technology, 51(9), 5279-5286 (2017)
J E Biles et al.
Journal of agricultural and food chemistry, 47(5), 1965-1969 (1999-12-20)
Migration of the diglycidyl ether of bisphenol A (DGEBA) to food from can enamels and can pull-top seals is reported. Derivatives of DGEBA are also determined in some foods. Levels of DGEBA in the foods surveyed in this study range
M. Biedermann et al.
Mitteilungen Aus Dem Gebiete Der Lebensmittel-Untersuchung Un Hygiene = Travaux De Chimie Alimentaire Et D'Hygiene, 90, 177-177 (1999)
Amira M El-Kosasy et al.
Journal of chromatographic science, 56(10), 920-932 (2018-07-10)
Gradient reversed-phase high-performance liquid chromatography with photodiode array detection was used for separation, detection and quantification of bisphenol-A-diglycidyl ether (BADGE) and some of its reaction products, namely, BADGE·HCl·H2O, BADGE·H2O and BADGE·2HCl in pure form and in canned foods, where canned
Natalia Szczepańska et al.
The Science of the total environment, 708, 135178-135178 (2019-12-04)
The problem of the presence of trace organic pollutants in food is of growing importance due to increasing awareness about their impact on newborns, infants and adults of reproductive age. Despite the fact that packaged food products offer many advantages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service